Skip to Content
Merck
2-Amino-N-cyclohexyl-N-methylbenzylamine 98%

2-Amino-N-cyclohexyl-N-methylbenzylamine

2-Amino-N-cyclohexyl-N-methylbenzylamine 98%
Synonym(s):
N-(2-Aminobenzyl)-N-methylcyclohexanamine
Linear Formula:
H2NC6H4CH2N(C6H11)CH3
CAS Number:
Molecular Weight:
218.34
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing