Skip to Content
Merck
2,3-Difluorobenzyl bromide 98%

2,3-Difluorobenzyl bromide

2,3-Difluorobenzyl bromide 98%
Synonym(s):
α-Bromo-2,3-difluorotoluene
Linear Formula:
F2C6H3CH2Br
CAS Number:
Molecular Weight:
207.02

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing