Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,2,3,3,3-Pentafluoro-1-propanol
Synonym(s):
PFPOH
Linear Formula:
CF
3
CF
2
CH
2
OH
CAS Number:
422-05-9
Molecular Weight:
150.05
EC Number:
207-012-7
Products
Product Category
building blocks (1)
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
257478
2,2,3,3,3-Pentafluoro-1-propanol,
97%
Expand
View Pricing
79879
2,2,3,3,3-Pentafluoro-1-propanol,
derivatization grade (GC derivatization), LiChropur
™
, ≥98.5% (GC)
Expand
View Pricing