Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,2,2-Trifluoroethanol
Synonym(s):
Trifluoroethyl alcohol, 2,2,2-Trifluoroethanol
Linear Formula:
CF
3
CH
2
OH
CAS Number:
75-89-8
Molecular Weight:
100.04
EC Number:
200-913-6
Products
Product Category
solvents (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
T63002
2,2,2-Trifluoroethanol,
ReagentPlus
®
, ≥99%
Expand
View Pricing
8.08259
2,2,2-Trifluoroethanol,
for synthesis
Expand
View Pricing
91683
2,2,2-Trifluoroethanol,
analytical standard, for NMR spectroscopy
Expand
View Pricing