Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1-Oleoyl-
rac
-glycerol
Synonym(s):
1-(
cis
-9-Octadecenoyl)-
rac
-glycerol,
rac
-Glycerol 1-monooleate, DL-α-Monoolein, Glyceryl
cis
-9-octadecenoate, Monoolein
Linear Formula:
CH
3
(CH
2
)
7
CH=CH(CH
2
)
7
COOCH
2
CHOHCH
2
OH
CAS Number:
111-03-5
Molecular Weight:
356.54
EC Number:
203-827-7
Products
Product Category
detergents (2)
glycerides (1)
lipids (1)
metabolites (1)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Technical (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
M7765
1-Oleoyl-
rac
-glycerol,
≥99%
Expand
View Pricing
49960
1-Oleoyl-
rac
-glycerol,
technical, ~40% (TLC)
Expand
View Pricing