Skip to Content
Merck
All Photos(1)

Documents

1508901

USP

Pentoxifylline

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,7-Dimethyl-1-(5-oxohexyl)xanthine, Trental

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18N4O3
CAS Number:
Molecular Weight:
278.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pentoxifylline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1C=NC2C1C(=O)N(CCCCC(C)=O)C(=O)N2C

InChI

1S/C13H20N4O3/c1-9(18)6-4-5-7-17-12(19)10-11(14-8-15(10)2)16(3)13(17)20/h8,10-11H,4-7H2,1-3H3

InChI key

MQGNNJQTNFHNHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pentoxifylline USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Pentoxifylline Compounded Oral Suspension
  • Pentoxifylline Extended-Release Tablets

Biochem/physiol Actions

Phosphodiesterase inhibitor; inhibits synthesis of tumor necrosis factor α (TNF-α).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Lact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Shan et al.
The Cochrane database of systematic reviews, 2(2), CD006800-CD006800 (2012-02-18)
Diabetic kidney disease (DKD) is associated with increased morbidity and mortality, mostly relating to cardiovascular complications. The relevance of inflammation in the pathogenesis of DKD has been investigated in recent years, and it has been shown that inflammatory markers are
Andrew B Jull et al.
The Cochrane database of systematic reviews, 12, CD001733-CD001733 (2012-12-14)
Healing of venous leg ulcers is improved by the use of compression bandaging but some venous ulcers remain unhealed, and some people are unsuitable for compression therapy. Pentoxifylline, a drug which helps blood flow, has been used to treat venous
R Parker et al.
Alimentary pharmacology & therapeutics, 37(9), 845-854 (2013-03-16)
Acute alcoholic hepatitis (AH) is a severe manifestation of alcoholic liver disease with a grave prognosis. Pentoxifylline, an oral antitumour necrosis factor agent, has been reported to reduce mortality and incidence of hepatorenal syndrome (HRS) in severe alcoholic hepatitis (SAH).
Donghao Lu et al.
The Cochrane database of systematic reviews, 1, CD007677-CD007677 (2012-01-20)
Endometriosis is a chronic, recurring condition that occurs during the reproductive years. It is characterized by endometrial tissue developing outside the uterine cavity. This endometrial tissue development is dependent on oestrogen produced primarily by the ovaries and, therefore, traditional management
Donghao Lv et al.
The Cochrane database of systematic reviews, (3)(3), CD007677-CD007677 (2009-07-10)
Endometriosis is a chronic, recurring condition that occurs during the reproductive years. It is characterized by endometrial tissue developing outside the uterine cavity. This endometrial tissue development is dependent on estrogen produced primarily by the ovaries and, therefore, traditional management

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service