Skip to Content
MilliporeSigma

Skip To

1181007

USP

Dextromethorphan hydrobromide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dextromethorphan hydrobromide monohydrate, (9S,13S,14S)-3-Methoxy-17-methylmorphinan hydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Please contact Customer Service for Availability
$621.00

About This Item

Empirical Formula (Hill Notation):
C18H25NO · HBr · H2O
CAS Number:
Molecular Weight:
370.32
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
6453793

$621.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

dextromethorphan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Br[H].[H]O[H].[H][C@]12CCCC[C@]13CCN(C)[C@H]2Cc4ccc(OC)cc34

InChI

1S/C18H25NO.BrH.H2O/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18;;/h6-7,12,15,17H,3-5,8-11H2,1-2H3;1H;1H2/t15-,17+,18+;;/m1../s1

InChI key

STTADZBLEUMJRG-IKNOHUQMSA-N

Gene Information

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextromethorphan hydrobromide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Acetaminophen, Chlorpheniramine Maleate, and Dextromethorphan Hydrobromide Tablets
  • Acetaminophen, Dextromethorphan Hydrobromide, Doxylamine Succinate, and Pseudoephedrine Hydrochloride Oral Solution
  • Capsules Containing at Least Three of the Following—Acetaminophen and Salts of Chlorpheniramine, Dextromethorphan, and Pseudoephedrine
  • Dextromethorphan Hydrobromide
  • Dextromethorphan Hydrobromide Oral Solution
  • Guaifenesin, Pseudoephedrine Hydrochloride, and Dextromethorphan Hydrobromide Capsules
  • Oral Powder Containing at Least Three of the Following—Acetaminophen and Salts of Chlorpheniramine, Dextromethorphan, and Pseudoephedrine

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D9684Y0002402D2531
grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical

application(s)

-

format

neat

format

-

format

-

format

-

API family

dextromethorphan

API family

-

API family

-

API family

-

Gene Information

human ... GRIN2A(2903), SIGMAR1(10280)

Gene Information

human ... GRIN2A(2903), SIGMAR1(10280)

Gene Information

-

Gene Information

human ... GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRIN3A(116443), GRIN3B(116444), SIGMAR1(10280)


Still not finding the right product?

Explore all of our products under Dextromethorphan hydrobromide


pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters