Skip to Content
Merck
All Photos(1)

Key Documents

89529

Supelco

Maleic anhydride

analytical standard

Synonym(s):

2,5-Furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2O3
CAS Number:
Molecular Weight:
98.06
Beilstein:
106909
MDL number:
UNSPSC Code:
12162002

grade

analytical standard

Quality Level

vapor density

3.4 (vs air)

vapor pressure

0.16 mmHg ( 20 °C)

autoignition temp.

870 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7.1 %

bp

200 °C (lit.)

mp

51-56 °C (lit.)

SMILES string

O=C1OC(=O)C=C1

InChI

1S/C4H2O3/c5-3-1-2-4(6)7-3/h1-2H

InChI key

FPYJFEHAWHCUMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1A - STOT RE 1 Inhalation

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service