Skip to Content
Merck
All Photos(1)

Key Documents

57610-U

Supelco

Rezorian A161 Cartridge

size 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724

form

prepacked column

packaging

pkg of 6 ea

technique(s)

LPLC: suitable

size

1 mL

surface area

800-900 m2/g

matrix

styrene-divinylbenzene, spherical, reagent grade

matrix active group

polymer

mean particle size

120 μm

pore size

110-175 Å pore diameter

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Disposable Rezorian A161 Luer lock syringe-tip cartridges offer convenience and expedience for isolating, purifying, and concentrating biomolecules from aqueous samples. Packed with a high performance, macroreticular, hydrophobic adsorbent resin, Rezorian A161 cartridges are specially tailored for biomolecular pharmaceutical separations.

Empty Rezorian tubes are available, and we may custom prepare Rezorian cartridges for your application - please inquire through Technical Service at [email protected].

Application

Rezorian A161 cartridge was used to perform dynamic column experiment to verify the direct separation of radionuclides from liquid decontamination waste and to determine Cu, Cd, Mo, As and Sb elements in biological samples.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of radionuclides from spent decontamination solutions onto selective inorganic-organic composite absorbers.
Rosikova, K., et al.
Czechoslovak Journal of Physics, 53, A603-A610 (2003)
Use of new composite materials for the determination of Cu, Cd, Mo, As, and Sb in biological samples by radiochemical neutron activation analysis.
Lucanikova, M., et al.
J. Radioanal. Nucl. Chem., 269, 463-468 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service