Skip to Content
Merck
All Photos(1)

Key Documents

442685

Supelco

N-Nitrosodi-n-butylamine

analytical standard

Synonym(s):

N-Nitrosodibutylamine, Dibutyl nitrosamine, NDBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O
CAS Number:
Molecular Weight:
158.24
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.9 g/mL (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CCCCN(CCCC)N=O

InChI

1S/C8H18N2O/c1-3-5-7-10(9-11)8-6-4-2/h3-8H2,1-2H3

InChI key

YGJHZCLPZAZIHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Nitrosodi-n-butylamines (NDBAs) are classified under the N-nitrosamine family of compounds which are considered as trace organic contaminants (TrOC) in drinking water and reclaimed effluent.

Application

NDBA has been used as a reference standard for the determination of the analyte in water by solid phase extraction (SPE) combined with gas chromatography-triple quadrupole mass spectrometry and in meat products by SPE combined with gas chromatography-nitrogen chemiluminescence detection (GC-NCD). It may be used as a reference standard for the analysis of residual NDBA in wastewater after reverse osmosis and UV treatment processes with SPE followed by liquid chromatography-tandem mass spectrometry (SPE-LC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new method for quantifying N-nitrosamines in wastewater samples by gas chromatography?triple quadrupole mass spectrometry
Yoon S, et al.
Talanta, 97, 256 -2261 (2012)
N-nitrosodimethylamine (NDMA) removal by reverse osmosis and UV treatment and analysis via LC -MS/MS
Plumlee MH, et al.
Water Research, 42(1-2), 347 -3355 (2008)
Determination of volatile nitrosamines in various meat products using comprehensive gas chromatography -nitrogen chemiluminescence detection
Ozel MZ, et al.
Food And Chemical Toxicology, 48(11), 3268 -33273 (2010)
Analysis of N-nitrosamines in water by isotope dilution gas chromatography -electron ionisation tandem mass spectrometry
McDonald JA, et al.
Talanta, 99, 146 -1154 (2012)
Mohamed F Ismail et al.
International journal of biological sciences, 5(4), 377-387 (2009-06-13)
Spirulina platensis (SP) is a filamentous cyanobacterium microalgae with potent dietary phyto-antioxidant, anti-inflammatory and anti-cancerous properties. The present study aimed to investigate the chemopreventive effect of SP against rat liver toxicity and carcinogenesis induced by dibutyl nitrosamine (DBN) precursors, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service