Skip to Content
Merck
All Photos(19)

Key Documents

ZRB1226

Sigma-Aldrich

Anti-phospho Histone H3 S10 Antibody, clone 63-1C-8 , ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

H3/g, H3t, Histone H3/t

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

63-1C-8, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 15.51 kDa
observed mol wt ~17 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

dot blot: suitable
flow cytometry: suitable
immunocytochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer10)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 63-1C-8 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Histone H3 phosphorylated on serine 10. It targets an epitope within 14 amino acids surrounding phosphoserine 10 from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids surrounding phosphoserine 10 of human Histone H3.

Application

Anti-phospho Histone H3 S10, clone 63-1C-8 ZooMAb, Cat. No. ZRB1226, is a recombinant rabbit monoclonal antibody that targets phospho-Histone H3 (Ser10) and has been tested for use in Dot Blot, Flow Cytometry, Immunocytochemistry, Peptide Inhibition Assay, and Western Blotting.
Flow Cytometry Analysis: 0.01 μg from a representative lot detected phospho-Histone H3 (Ser10) in one million Jurkat cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-Histone H3 (Ser10) in HeLa, A431, HUVEC, and NIH 3T3 cells.

Dot Blot Analysis: Representative lot was tested on AbSurance Histone Peptide Microarray.


Peptide Inhibition Assay Analysis: 1:100 dilution from a representative lot did not detect the target band in acid extract from HeLa cells treated with colemid (50 ng/ml) when preblocked with the S10 phosphopeptide, but detected the band when the corresponding non-phosphopeptide was used.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Histone H3.1t (UniProt: Q16695; also known as H3/t, H3t, H3/g) is encoded by the HIST3H3 (also known as H3FT) gene (Gene ID: 8290) in human. Histone H3 has two main variants, H3.1 and H3.3, which show different genomic localization patterns in animals. The H3.1 and H3.3 complexes also possess distinct histone chaperones, CAF-1 and HIRA, which play important role in mediating DNA-synthesis-dependent and -independent nucleosome assembly. It has been reported that Histone H3.1 serves as the canonical histone, which is incorporated during DNA replication, whereas H3.3 acts as the replacement histone that can be incorporated outside of S-phase during chromatin-disrupting processes like transcription. Histone H 3.1 is a core component of nucleosome that is present only in mammals and is usually enriched in acetylation of lysine 14 and demethylation of lysine 9 (with initial methionine removed; HeK9Me2). It is expressed during S phase and its expression decreases significantly as cell division slows down during the process of differentiation. Phosphorylation of serine 10 (with initiator methionine removed) in the tails of histone H3 is an important modification that is involved in both transcription and cell division. This phosphorylation is mainly achieved by the action signaling as well as mitotic kinases. This phosphorylation is reported to be dependent on the post-translational modification of neighboring amino acids. A reduction in methylation of lysine 9 can increase phosphorylation of serine 10, but demethylation of lysine 9 reduces serine 10 phosphorylation. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Prigent, C., and Dimitrov, S. (2003). J. Cell Sci. 116(18); 3677-3685; Stroud, H., et al (2012). Proc. Natl. Acad. Sci. USA 109(14); 5370-5375).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Divya A Shiroor et al.
EMBO reports, 24(5), e56112-e56112 (2023-03-22)
As stem cells divide, they acquire mutations that can be passed on to daughter cells. To mitigate potentially deleterious outcomes, cells activate the DNA damage response (DDR) network, which governs several cellular outcomes following DNA damage, including repairing DNA or

Protocols

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Related Content

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

Greener ZooMAb® antibodies offer sustainable options with low-waste packaging and BSA-free formulation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service