Skip to Content
Merck
All Photos(20)

Key Documents

ZMS1023

Sigma-Aldrich

Anti-Rac1 Antibody, clone 23A8 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cell migration-inducing gene 5 protein, EC:3.6.5.2, Ras-like protein TC25, Ras-related C3 botulinum toxin substrate 1, p21-Rac1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

23A8, recombinant monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 21.45 kDa
observed mol wt ~20 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 23A8 is a ZooMAb mouse recombinant monoclonal antibody that specifically detects RAC1.

Immunogen

Full-length recombinant human RAC1.

Application

Quality Control Testing

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Rac1 in Rat Brain Microsomes.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Rac1 in A431, HeLa, and NIH3T3 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 silution from a representative lot detected Rac1 in human esophagus tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Rac1 in HeLa, A431, HUVEC , and NIH 3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Rac1, clone 23A8 ZooMAb, Cat. No. ZMS1023, is a recombinant Mouse monoclonal antibody that specifically targets Rac1 and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Ras-related C3 botulinum toxin substrate 1 (UniProt: P63000; also known as EC:3.6.5.2, Cell migration-inducing gene 5 protein, Ras-like protein TC25, p21-Rac1) is encoded by the RAC1 (also known as TC25, MIG5) gene (Gene ID: 5879) in human. Rac1 is a plasma membrane-associated small GTPase that cycles between active GTP-bound and inactive GDP-bound states. In its active state, it binds to a variety of effector proteins to regulate cellular responses. Two isoforms of Rac1 have been described that are produced by alternative splicing. Rac1b differs from Rac1a in that it has an additional 19 amino acids and is found in various tumors, such as breasts and colorectal. Rac1b has an accelerated GEF-independent GDP/GTP exchange and an impaired GTP hydrolysis, which is restored partially by GTPase-activating proteins. It can bind to the GTPase-binding domain of PAK, but not full-length PAK in a GTP-dependent manner, suggesting that the insertion does not completely abolish effector interaction. Isoform B is predominantly identified in skin and epithelial tissues from the intestinal tract. Its expression is elevated in colorectal tumors at various stages of neoplastic progression, as compared to their respective adjacent tissues. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service