The plastic base of this plate is full of fluorescent particles, creating an enormous amount of uneven background signal in all visible wavelengths. I inquired if this was just a defect with my plate but apparently this "has been the case for years". They said it was possible to cut out this fluorescence with confocal microscopy but the fibers are only a couple of micrometers from the base so it will still be visible in most cases. This product is entirely unsuitable for any sort of fluorescence microscopy and the partially opaque fibers make it unuseable for brightfield as well, despite being advertised as a tool for "live cell imaging" and similar applications.
Z694541
Nanofiber multiwell plate, with aligned nanofibers
size 96 wells
Synonym(s):
3D, 3D Cell Culture, 3d plates, 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates, tissue culture plates
About This Item
flat bottom clear wells
transparent polycaprolactone (PCL)
Recommended Products
material
(polycaprolactone (PCL))
flat bottom clear wells
transparent polycaprolactone (PCL)
sterility
sterile
feature
lid
manufacturer/tradename
Nanofiber Solutions 9602
size
96 wells
suitability
suitable for (Tissue Engineering, Cancer Research)
binding type
non-treated surface
General description
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
For fluorescent imaging applications, we recommend using the Nanofiber Solutions chamber slides.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Articles
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models