Skip to Content
Merck
All Photos(2)

Key Documents

WH0006925M2

Sigma-Aldrich

Monoclonal Anti-TCF4 antibody produced in mouse

clone 1F6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-E22, Anti-ITF2, Anti-SEF2, Anti-SEF21, Anti-SEF21A, Anti-SEF21B, Anti-transcription factor 4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TCF4(6925)

General description

Transcription factor 4 (TCF4), also known as E2-2, is a helix-loop-helix transcription factor. It is part of the E protein family and is expressed in the cornea during development. The gene encoding TCF4 is localized on human chromosome 18q21.

Immunogen

TCF4 (AAH31056.1, 1 a.a. ~ 365 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MHHQQRMAALGTDKELSDLLDFSAMFSPPVSSGKNGPTSLASGHFTGSNVEDRSSSGSWGNGGHPSPSRNYGDGTPYDHTTSRDLGSHDNLSPPFVNSRIQSKTERGSYSSYGRESNLQGCHQQSLLGGDMDMGNPGTLSPTKPGSQYYQYSSNNPRRRPLHSSAMEVQTKKVRKVPPGLPSSVYAPSASTADYNRDSPGYPSSKPATSTFPSSFFMQDGHHSSDPWSSSSGMNQPGYAGMLGNSSHIPQSSSYCSLHPHERLSYPSHSSADINSSLPPMSTFHRSGTNHYSTSSCTPPANGTDSIMANRGSGAAGSSQTGDALGKALASIYSPDHTNNSFSSNPSTPVGSPPSLSAGTAVWSRN

Biochem/physiol Actions

Transcription factor 4 (TCF4) acts a regulator of cellular growth and differentiation. It has a role in the development of central nervous system. Defects in TCF4 gene are associated with Pitt-Hopkins syndrome. The protein has also been linked to susceptibility to schizophrenia.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TCF4 deletions in Pitt-Hopkins Syndrome.
Giurgea L
Human Mutation, 29(11), E242-E251 (2008)
TGC repeat expansion in the TCF4 gene increases the risk of Fuchs' endothelial corneal dystrophy in Australian cases.
Kuot A
PLoS ONE, 12(8), e0183719-e0183719 (2017)
Association of TCF4 polymorphisms and Fuchs' endothelial dystrophy: a meta-analysis.
Li D
BMC Ophthalmology, 15, 61-61 (2015)
Complex translocation disrupting TCF4 and altering TCF4 isoform expression segregates as mild autosomal dominant intellectual disability.
Maduro V
Orphanet Journal of Rare Diseases, 11(1), 62-62 (2016)
TCF4 gene polymorphism is associated with cognition in patients with schizophrenia and healthy controls.
Journal of Psychiatric Research, 95-101 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service