Skip to Content
Merck
All Photos(6)

Key Documents

WH0004435M1

Sigma-Aldrich

Monoclonal Anti-CITED1 antibody produced in mouse

clone 6G8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1, Anti-MSG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G8, monoclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CITED1(4435)

General description

CITED1 (Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1)/MSG1 (melanocyte-specific gene 1) is a non-DNA binding transcriptional co-activator. It has a smad-4 interacting domain (SID) and a conserved region 2 (CR2) domain. This gene codes for a 27 kDa protein that belongs to the CITED (CBP/p300-interacting transactivators with glutamic acid [E] and aspartic acid [D]-rich C-terminal domain) family of nuclear proteins. CITED1 is located on human chromosome Xq13.
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described. (provided by RefSeq)

Immunogen

CITED1 (NP_004134, 94 a.a. ~ 193 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SMQLQKLNSQYQGMAAATPGQPGEAGPLQNWDFGAQAGGAESLSPSAGAQSPAIIDSDPVDEEVLMSLVVELGLDRANELPELWLGQNEFDFTADFPSSC

Biochem/physiol Actions

CITED1 (Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1)/MSG1 (melanocyte-specific gene 1) controls cellular activities of the metanephric mesenchyme. This gene is linked to papillary thyroid carcinoma. MSG1 is involved in pigmentation. It also participates in malignant transformation of pigment cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wilms' tumorigenesis is altered by misexpression of the transcriptional co-activator, CITED1
Lovvorn HN 3rd, et al.
Journal of Pediatric Surgery, 42(3), 474-481 (2007)
Establishment and genetic characterization of a novel mixed-phenotype acute leukemia cell line with EP300-ZNF384 fusion
Ping N, et al.
Journal of Hematology & Oncology, 8(1), 100-100 (2015)
Aberrant expression of MSG1 transcriptional activator in human malignant melanoma in vivo
Ahmed NU, et al.
Pigment Cell Research / Sponsored by the European Society for Pigment Cell Research and the International Pigment Cell Society, 14(2), 140-143 (2001)
Galectin-3, fibronectin-1, CITED-1, HBME1 and cytokeratin-19 immunohistochemistry is useful for the differential diagnosis of thyroid tumors
Prasad ML, et al.
Modern Pathology, 18(1), 48-57 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service