Skip to Content
Merck
All Photos(1)

Key Documents

T8778

Sigma-Aldrich

Trehalase from porcine kidney

buffered aqueous glycerol solution, ≥1.0 units/mg protein

Synonym(s):

α,α-Trehalose glucohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

buffered aqueous glycerol solution

Quality Level

specific activity

≥1.0 units/mg protein

concentration

0.5-10.0 mg/mL protein (BCA)

storage temp.

−20°C

General description

Trehalase is a glycosidase that occurs in many forms and is found in all tissues. It belongs to the glycoside hydrolase 37 (GH37) family of the carbohydrate-active enzyme (CAZy) classification.
Trehalose is the main haemolymph sugar in most insects and is potentially a prime target for an invading pathogenic fungus.

Application

Trehalase has been used:
  • in a study to assess changes in carbohydrate metabolism in Plasmopara viticola-infected grapevine leaves
  • in a study to investigate growth arrest by trehalose-6-phosphate
  • to quantify the intracellular trehalose in Lactobacillus casei cells

Biochem/physiol Actions

Trehalase catalyzes the hydrolysis of trehalose to produce two glucose units. It mediates flight metabolism, molting induced chitin production and tolerance towards cold temperature.

Unit Definition

One unit will convert 1.0 μmole of trehalose to 2.0 μmoles of glucose per min at pH 5.7 at 37 °C (liberated glucose determined at pH 7.5).

Physical form

Solution in 50% glycerol containing 1% Triton X-100 and 25 mM potassium phosphate, pH 6.5

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shari Dhaene et al.
Journal of enzyme inhibition and medicinal chemistry, 35(1), 1964-1989 (2020-11-10)
Although trehalose has recently gained interest because of its pharmaceutical potential, its clinical use is hampered due to its low bioavailability. Hence, hydrolysis-resistant trehalose analogues retaining biological activity could be of interest. In this study, 34 4- and 6-O-substituted trehalose
Growth arrest by trehalose-6-phosphate: an astonishing case of primary metabolite control over growth by way of the SnRK1 signaling pathway
Delatte, T., et al.
Annual review of plant physiology and plant molecular biology, 157, 160-174 (2011)
Berrak Gülçin Balaban et al.
Microorganisms, 8(1) (2019-12-28)
Iron plays an essential role in all organisms and is involved in the structure of many biomolecules. It also regulates the Fenton reaction where highly reactive hydroxyl radicals occur. Iron is also important for microbial biodiversity, health and nutrition. Excessive
Christopher Askew et al.
PLoS pathogens, 5(10), e1000612-e1000612 (2009-10-10)
Glycolysis is a metabolic pathway that is central to the assimilation of carbon for either respiration or fermentation and therefore is critical for the growth of all organisms. Consequently, glycolytic transcriptional regulation is important for the metabolic flexibility of pathogens
Jiangnan Luo et al.
PloS one, 9(6), e99732-e99732 (2014-06-14)
A set of 14 insulin-producing cells (IPCs) in the Drosophila brain produces three insulin-like peptides (DILP2, 3 and 5). Activity in IPCs and release of DILPs is nutrient dependent and controlled by multiple factors such as fat body-derived proteins, neurotransmitters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service