Skip to Content
MilliporeSigma

Skip To

SML2600

Melitracen hydrochloride

≥98% (HPLC)

Synonym(s):

Melitracen hydrochloride, 3-(10,10-dimethyl-9(10H)-anthracenylidene)-N,N-dimethyl-1-propanamine hydrochloride, U-24973A hydrochloride, 3-[10,10-Dimethyl-9(10H)-anthrylidene]-N,N-dimethylpropylamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg

Available to ship TODAYfromMILWAUKEE

$91.10
$77.43

About This Item

Empirical Formula (Hill Notation):
C21H25N · HCl
CAS Number:
Molecular Weight:
327.89
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

$77.43

List Price$91.10Save 15%
Web-Only Promotion

Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[Cl-].[N+H](CCC=C1c2c(cccc2)C(c3c1cccc3)(C)C)(C)C

InChI

1S/C21H25N.ClH/c1-21(2)19-13-7-5-10-17(19)16(12-9-15-22(3)4)18-11-6-8-14-20(18)21;/h5-8,10-14H,9,15H2,1-4H3;1H

InChI key

RADLXCPDUXFGFF-UHFFFAOYSA-N

Biochem/physiol Actions

Melitracen is a tricyclic antidepressant (TCA) that likely exerts its in vivo efficacy similarly as imipramine and amitriptyline by inhibiting serotonin-norepinephrine reuptake.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP1151SML0190SML2570
assay

≥98% (HPLC)

assay

-

assay

≥95% (HPLC)

assay

≥98% (HPLC)

form

powder

form

solid

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: 2 mg/mL, clear (warmed)

solubility

-

solubility

DMSO: ≥10 mg/mL

solubility

DMSO: 2 mg/mL, clear

storage condition

desiccated

storage condition

-

storage condition

desiccated

storage condition

desiccated

color

white to beige

color

-

color

white to tan

color

white to beige


Still not finding the right product?

Explore all of our products under Melitracen hydrochloride


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters