Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C11H8O4
CAS Number:
Molecular Weight:
204.18
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Product Name
4μ8C, ≥98% (HPLC)
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 5 mg/mL, clear (warmed)
storage temp.
−20°C
SMILES string
O1c2c(ccc(c2C=O)O)C(=CC1=O)C
InChI
1S/C11H8O4/c1-6-4-10(14)15-11-7(6)2-3-9(13)8(11)5-12/h2-5,13H,1H3
InChI key
RTHHSXOVIJWFQP-UHFFFAOYSA-N
Biochem/physiol Actions
4μ8C is a potent inhibitor of the ER transmembrane protein IRE1, which mediates the unfolded protein response. 4μ8C inhibits regulated IRE1-dependent degradation (RIDD) and unconventional spicing of XBP1 mRNA in response to endopasmic reticulum (ER) stress, but has no affect on the kinase activity of IRE1.
4μ8C is a potent inhibitor of the ER transmembrane protein IRE1.
4μ8C is a salicylaldehyde derivative and its inhibitory action targets the K907 lysine residue in RNase (ribonuclease) domain and affects IRE1α (endoplasmic reticulum to nucleus signaling 1) RNase activity. In vitro analysis proved that 4μ8C was able to inhibit the release of insulin at high glucose levels.
Features and Benefits
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| form powder | form powder | form powder | form powder |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
| solubility DMSO: 5 mg/mL, clear (warmed) | solubility DMSO: 1 mg/mL, clear (warmed) | solubility DMSO: 5 mg/mL, clear (warmed) | solubility DMSO: 5 mg/mL, clear (warmed) |
| color white to beige | color white to light brown | color white to beige | color white to beige |
Still not finding the right product?
Explore all of our products under 4μ8C
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



