Skip to Content
Merck
All Photos(1)

Key Documents

S9572

Sigma-Aldrich

Anti-Sonic Hedgehog peptide (N-terminal) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Dsh, Anti-Hhg1, Anti-Hx, Anti-Shh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

immunohistochemistry: 15 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Shh(20423)

General description

The Hedgehog (Hh) family of secreted proteins consists of sonic hedgehog (Shh), desert hedgehog (Dhh) and Indian hedgehog (Ihh). The Hh signaling regulates cell migration and tissue construction during embryogenesis and reconstruction in adults. Shh protein is secreted by Purkinje cells in the cerebellum with both autocrine and paracrine functions. The Shh signaling is mediated by the interaction of Shh with transmembrane proteins, Patched and Smoothened. The Shh pathway cross-talks Notch and Wnt pathways and acts via downstream targets such as cyclins, γ catenin, myc and other transcription factors. The Hh family members have multiple roles as morphogens, cell survival factors, setting anterior-posterior axis and digit formation during development of limb and axon guidance factors during the development of brain and nervous system. Hedgehog signaling has been implicated in childhood tumors of pancreas, liver, thyroid, gastrointestinal tract, neuroblastomas and medulloblastomas
Anti-Sonic Hedgehog Peptide (N-terminal) recognizes mouse Sonic Hedgehog N-terminal peptide.

Immunogen

purified, E. coli-derived recombinant mouse 6× histidine-tagged sonic hedgehog N-terminal peptide.

Application

Anti-Sonic Hedgehog Peptide (N-terminal) can be used for immunoblotting at a working concentration of 0.1-0.2 μg/ml. For immunohistochemistry, a working concentration of 15 μg/ml is recommended.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of Sonic hedgehog signaling pathway in neuroblastoma development
Shahi MH et al
Biol. Med., 1, Rev 2-Rev 2 (2009)
X Wu et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(1), G295-G306 (2007-11-23)
Saccharomyces boulardii has received increasing attention as a probiotic effective in the prevention and treatment of infectious and inflammatory bowel diseases. The aim of this study was to examine the ameliorating effects of S. boulardii on Citrobacter rodentium colitis in
Gijs R van den Brink
Physiological reviews, 87(4), 1343-1375 (2007-10-12)
The Hedgehog family of secreted morphogenetic proteins acts through a complex evolutionary conserved signaling pathway to regulate patterning events during development and in the adult organism. In this review I discuss the role of Hedgehog signaling in the development, postnatal
Fergal C Kelleher
Carcinogenesis, 32(4), 445-451 (2010-12-28)
To conduct a systematic review of the role that the hedgehog signaling pathway has in pancreatic cancer tumorigenesis. PubMed search (2000-2010) and literature based references. Firstly, in 2009 a genetic analysis of pancreatic cancers found that a core set of
Shohreh F Farzan et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(4), G844-G849 (2008-02-02)
The Hedgehog (Hh) family of secreted proteins is necessary for aspects of the development and maintenance of the gastrointestinal tract. Hh is thought to function as a morphogen, a mitogen, a cell survival factor, and an axon guidance factor. Given

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service