Skip to Content
Merck
All Photos(1)

Key Documents

S9399

Sigma-Aldrich

Staphylococcal enterotoxin A

from Staphylococcus aureus, lyophilized powder, enterotoxin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Staphylococcal enterotoxin A from Staphylococcus aureus,

contains

sodium phosphate buffer as balance

Quality Level

composition

Protein, ~10% Lowry

availability

not available in USA

impurities

Staphylococcal enterotoxin B (SEB), essentially free

solubility

H2O: complete 1 mg/ml, clear, colorless

storage temp.

2-8°C

General description

Staphylococcal enterotoxin A belongs to the family of staphylococcal exotoxins. It is secreted by Staphylococcus aureus, a Gram-positive bacteria.

Application

Staphylococcal enterotoxin A from Staphylococcal aureus was injected peritoneally in mice to induce sepsis.10
Staphylococcal enterotoxin A from Staphylococcus aureus has been used as an external standard in for the verification of TRANSIA assay kit, RIDASCREEN assay kit and Vidas Set 2 method. For in vitro toxin stimulation of mouse splenocytes.

Biochem/physiol Actions

Staphylococcal enterotoxin A from Staphylococcus aureus is leading agent that causes of food poisoning. A concentration of as little as 0.5 mg/ml is sufficient to result in nausea, vomiting, diarrhea and cramps.8 It stimulates the cell proliferation of peripheral lymphocytes, induces the production of interferons and is important for gut immunity against S. aureus infections.9 It acts as a super antigen for T-lymphocytes.

Packaging

Package size based on protein content

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of staphylococcal enterotoxins in cheese by immunoenzyme assays
<BIG>Jankovic V, et al.</BIG>
Archives of Biological Sciences, 64, 1449-1454 (2012)
Marcin Kłak et al.
Scandinavian journal of infectious diseases, 42(5), 351-358 (2010-01-27)
Haemostatic balance shifts towards pro-coagulation during infection. Plasminogen, a key molecule of fibrinolysis, may play an important role in the pathogenesis of staphylococcal infections. In the present study, we assessed the impact of inhibition of plasminogen activation by tranexamic acid
Staphylococcal enterotoxin A (SEA).
L Spero et al.
Methods in enzymology, 78(Pt A), 331-336 (1981-01-01)
Hongshi Yu et al.
BMC genomics, 10, 416-416 (2009-09-08)
WNT4 is a critical signalling molecule in embryogenesis and homeostasis, but the elements that control its transcriptional regulation are largely unknown. This study uses comparative cross species sequence and functional analyses between humans and a marsupial (the tammar wallaby,Macropus eugenii)
Peter W Hunt et al.
PloS one, 6(1), e15924-e15924 (2011-02-10)
HIV-infected individuals maintaining undetectable viremia in the absence of therapy (HIV controllers) often maintain high HIV-specific T cell responses, which has spurred the development of vaccines eliciting HIV-specific T cell responses. However, controllers also often have abnormally high T cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service