Skip to Content
Merck
All Photos(2)

Documents

S3131

Sigma-Aldrich

Sulindac sulfide

≥98% (HPLC), solid

Synonym(s):

(Z)-5-Fluoro-2-methyl-1-[p-(methylthio)benzylidene]indene-3-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17FO2S
CAS Number:
Molecular Weight:
340.41
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

DMSO: ≥22 mg/mL

SMILES string

[H]\C(c1ccc(SC)cc1)=C2/C(C)=C(CC(O)=O)c3cc(F)ccc23

InChI

1S/C20H17FO2S/c1-12-17(9-13-3-6-15(24-2)7-4-13)16-8-5-14(21)10-19(16)18(12)11-20(22)23/h3-10H,11H2,1-2H3,(H,22,23)/b17-9-

InChI key

LFWHFZJPXXOYNR-MFOYZWKCSA-N

Application

Human endothelial cells, HMEC-1 were treated with Sulindac sulfide and the effect on cell survival was studies by MTT assay.

Biochem/physiol Actions

Sulindac sulfide is a non-steroidal anti-inflammatory compound with a preference for COX-1; it is an inhibitor of Ras activation of Raf-1. It impairs nucleotide exchange on Ras by CDC25 and accelerates Ras hydrolysis of GTP by p120GAP. It is an active metabolite of sulindac. It is also shown to inhibit growth and induce apoptosis in human prostate cancer cells through a COX-1 and COX-2 independent mechanism. Sulindac sulfide is an analgesic that has antiproliferative and apoptotic effects. It inhibits the expression and activity of cyclooxygenase-2 in human colon cancer cells and reduces tumor burden in adenomatous polyposis patients.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Z Zhang et al.
Gastroenterology, 118(6), 1012-1017 (2000-06-02)
Many reports indicate that nonsteroidal anti-inflammatory drugs (NSAIDs) have antineoplastic effects, but the precise molecular mechanism(s) responsible are unclear. We evaluated the effect of cyclooxygenase (COX) inhibitors (NSAIDs) on human colon carcinoma cells (HCA-7) and identified several genes that are
C Herrmann et al.
Oncogene, 17(14), 1769-1776 (1998-10-20)
The non-steroidal anti-inflammatory drug sulindac is used in cancer prevention and therapy, but the molecular aspects of its anti-tumor effect remain unresolved. In vivo the prodrug sulindac, is converted into the metabolite sulindac sulfide. We found that sulindac sulfide strongly
Luise Richter et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(33), 14597-14602 (2010-08-04)
Following ectodomain shedding by beta-secretase, successive proteolytic cleavages within the transmembrane sequence (TMS) of the amyloid precursor protein (APP) catalyzed by gamma-secretase result in the release of amyloid-beta (Abeta) peptides of variable length. Abeta peptides with 42 amino acids appear
J T Lim et al.
Biochemical pharmacology, 58(7), 1097-1107 (1999-09-14)
We examined the activity of two metabolites of sulindac (a nonsteroidal anti-inflammatory drug), sulindac sulfide and sulindac sulfone (exisulind, Prevatec), and a novel highly potent analog of exisulind (CP248) on a series of human prostate epithelial cell lines. Marked growth
Tomas Borgegard et al.
The Journal of biological chemistry, 287(15), 11810-11819 (2012-02-16)
γ-Secretase-mediated cleavage of amyloid precursor protein (APP) results in the production of Alzheimer disease-related amyloid-β (Aβ) peptides. The Aβ42 peptide in particular plays a pivotal role in Alzheimer disease pathogenesis and represents a major drug target. Several γ-secretase modulators (GSMs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service