RAB0389
Human NT-3 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type urine
sample type plasma
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 4.12-3000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... NTF3(4908)
General description
The Human NT-3 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human NT-3 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human NT3
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mou Gao et al.
Experimental and therapeutic medicine, 13(3), 885-890 (2017-04-30)
Intracerebral neural stem cell (NSC) transplantation is beneficial for delivering stem cell grafts effectively, however, this approach may subsequently result in brain injury and secondary inflammation. To reduce the risk of promoting brain injury and secondary inflammation, two methods were
Alexandra Koshkina et al.
Nutrients, 11(8) (2019-07-31)
The purpose of this study was to explore the antidepressant-like effects of vitamin D3 at different doses (1.0, 2.5, and 5.0 mg/kg sc) on a model of depression produced by chronic unpredictable mild stress (CUMS) for 28 days in long-term
Zi-Xing Xu et al.
Artificial cells, nanomedicine, and biotechnology, 48(1), 362-376 (2020-01-05)
Microvascular disturbance, excessive inflammation and gliosis are key pathophysiologic changes in relation to functional status following the traumatic spinal cord injury (SCI). Continuous release of vascular endothelial growth factor (VEGF) to the lesion site was proved be able to promote
Zehra Betül Ahi et al.
Biomedical materials (Bristol, England), 15(5), 055024-055024 (2020-05-13)
Spinal cord injury is a devastating condition of the central nervous system, in which traditional treatments are largely ineffective due to the complex nature of the injured tissue. Therefore, biomaterial-based systems have been developed as possible alternative strategies to repair
Renata Cristiane Dos Reis et al.
Behavioural pharmacology, 31(1), 27-33 (2019-10-03)
It has been proposed that neurotrophin-3 acts in a manner that is opposed to nerve growth factor, especially in the modulation of heat hyperalgesia. Injury to the constriction of the infraorbital nerve (CION) is a well-established model of trigeminal neuropathic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service