Skip to Content
Merck
All Photos(2)

Key Documents

R9644

Sigma-Aldrich

Ribavirin

antiviral

Synonym(s):

1-β-D-Ribofuranosyl-1,2,4-triazole-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4O5
CAS Number:
Molecular Weight:
244.20
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

water: 19.60-20.40 mg/mL, clear, colorless to faintly yellow

antibiotic activity spectrum

viruses

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

NC(=O)c1ncn(n1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

InChI

1S/C8H12N4O5/c9-6(16)7-10-2-12(11-7)8-5(15)4(14)3(1-13)17-8/h2-5,8,13-15H,1H2,(H2,9,16)/t3-,4-,5-,8-/m1/s1

InChI key

IWUCXVSUMQZMFG-AFCXAGJDSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.
Chemical structure: nucleoside

Biochem/physiol Actions

Antiviral agent used against a wide variety of human viral infections, in particular, chronic hepatitis C, HIV, and adenovirus. Its metabolite, ribavirin 5′-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase, but many other mechanisms of action are also supported with experimental evidence.
Antiviral agent used against a wide variety of human viral infections, in particular, chronic hepatitis C, HIV, and adenovirus. Its metabolite, ribavirin 5′-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase.
Antiviral agent. Its metabolite, ribavirin 5′-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase. Used to inhibit purine (GTP pools) biosynthesis at the level of inosine monophosphate (IMP) dehydrogenase and as a precursor to the di- and tri-phosphates which inhibit viral RNA-dependent RNA polymerases.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Hua Liu et al.
Annals of internal medicine, 159(11), 729-738 (2013-12-04)
Data are limited on the efficacy and safety of pegylated interferon plus ribavirin for patients with hepatitis C virus genotype 1 (HCV-1) receiving hemodialysis. To compare the efficacy and safety of combination therapy with pegylated interferon plus low-dose ribavirin and
Fred Poordad et al.
The New England journal of medicine, 370(21), 1973-1982 (2014-04-15)
Interferon-containing regimens for the treatment of hepatitis C virus (HCV) infection are associated with increased toxic effects in patients who also have cirrhosis. We evaluated the interferon-free combination of the protease inhibitor ABT-450 with ritonavir (ABT-450/r), the NS5A inhibitor ombitasvir
M Colombo et al.
Gut, 63(7), 1150-1158 (2013-11-10)
Severe adverse events (AEs) compromise the outcome of direct antiviral agent-based treatment in patients with advanced liver fibrosis due to HCV infection. HEP3002 is an ongoing multinational programme to evaluate safety and efficacy of telaprevir (TVR) plus pegylated-interferon-α (PEG-IFNα) and
Marie Pellerin et al.
Viruses, 13(3) (2021-04-04)
Hepatitis E virus (HEV) is considered as an emerging global health problem. In most cases, hepatitis E is a self-limiting disease and the virus is cleared spontaneously without the need of antiviral therapy. However, immunocompromised individuals can develop chronic infection
Tao Meng et al.
Journal of virology, 88(10), 5803-5815 (2014-03-14)
In a screen for ribavirin resistance, a novel high-fidelity variant of human enterovirus 71 (EV71) with the single amino acid change L123F in its RNA-dependent RNA polymerase (RdRp or 3D) was identified. Based on the crystal structure of EV71 RdRp

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service