Skip to Content
Merck
All Photos(2)

Key Documents

R1905

Sigma-Aldrich

Anti-RACK1 (N-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-GNB2L1, Anti-Gnb2-rs1, Anti-Guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1, Anti-H12.3, Anti-HLC-7, Anti-Lung cancer oncogene 7, Anti-PIG21, Anti-Proliferation-inducing gene 21, Anti-Receptor of Activated Protein Kinase C1, Anti-protein homologous to chicken B complex protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

human, rat, mouse

technique(s)

immunoprecipitation (IP): 5-10 μL using HEK-293T cell lysates
indirect immunofluorescence: 1:100-1:200 using paraformaldehyde-fixed HeLa cells
western blot: 1:500-1:1,000 using HEK-293T cell lystes

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GNB2L1(10399)
mouse ... Gnb2l1(14694)
rat ... Gnb2l1(83427)

Related Categories

General description

Receptors for activated C-kinase (RACKs) are scaffold protein for Protein kinase C (PKC). It is composed of seven WD (Trp-Asp) repeats, thus resembling the structure of the subunit of G proteins.

Specificity

Anti-RACK1 (N-terminal) is produced in rabbit and specifically recognizes human RACK1. Anti-RACK1 (N-terminal) antibody is specific for human RACK1 (approx. 30 kDa). The antibody also reacts with rat and mouse RACK1. In immunoblotting, staining of the RACK1 band is specifically inhibited by the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 2-16 of human RACK1, conjugated to KLH via an C-terminal added cysteine residue. The corresponding sequence is conserved in human, rat and mouse.

Application

Anti-RACK1 (N-terminal) antibody is suitable for use in indirect immunofluorescence (1:100-1:200 using paraformaldehyde-fixed HeLa cells), western blot (1:500-1:1,000 using HEK-293T cell lysates) and immunoprecipitation (5-10 μL using HEK-293T cell lysates).
It is also used in immunoblotting.

Biochem/physiol Actions

RACK1 is a receptor for activated C kinases that functions downstream of the growth factor receptor signaling pathway. RACK1 also modulates several functions such as cell adhesion, chemotaxis and protrusion. Increased RACK1 expression has been associated with cell migration and metastasis in breast carcinoma. Thus, RACK1 can be used as a prognostic indicator for breast cancer
Receptors for activated C-kinase (RACKs) anchor activated Protein kinase C (PKC) at the site of translocation. Receptors for activated C-kinase 1 (RACK1) is the selective RACK for PKC-βII. RACK1 anchors other signaling enzymes such as Sarcoma (Src) tyrosine kinase, integrin and phosphodiesterase 4D5. The ability of RACK1 to interact simultaneously with different signaling molecules, allows it to integrate inputs from distinct signaling pathways. Accumulating evidence attributes to it the role of regulating several major nervous system pathways. RACK1 is a ribosomal protein and can make direct contact with ribosomal RNA. It recruits activated PKC to the ribosome, which in turn, phosphorylates initiation factor 6, leading to ribosome assembly.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RACK1, A multifaceted scaffolding protein: Structure and function
Adams D R, et al.
Cell communication and signaling : CCS, 9(1), 22-22 (2011)
Receptor for activated C kinase 1 (RACK1) and Src regulate the tyrosine phosphorylation and function of the androgen receptor
Kraus S, et al.
Cancer research, 66(22), 11047-11054 (2006)
Elisabeth A Cox et al.
Molecular biology of the cell, 14(2), 658-669 (2003-02-18)
Mammalian cDNA expression cloning was used to identify novel regulators of integrin-mediated cell-substratum adhesions. Using a focal adhesion morphology screen, we identified a cDNA with homology to a receptor for activated protein kinase C (RACK1) that induced a loss of
Fengjiao Liu et al.
CNS neuroscience & therapeutics, 27(12), 1518-1530 (2021-09-05)
Medulloblastoma (MB) is a malignant pediatric brain tumor that arises in the cerebellar granular neurons. Sonic Hedgehog subtype of MB (SHH-MB) is one of the major subtypes of MB in the clinic. However, the molecular mechanisms underlying MB tumorigenesis are
Betty Y Chang et al.
Oncogene, 21(50), 7619-7629 (2002-10-26)
RACK1 is one of a group of PKC-interacting proteins collectively called RACKs (Receptors for Activated C-Kinases). Previously, we showed that RACK1 also interacts with the Src tyrosine kinase, and is an inhibitor of Src activity and cell growth. PKC activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service