Skip to Content
Merck
All Photos(1)

Key Documents

PZ0019

Sigma-Aldrich

CP-945,598

≥98% (HPLC)

Synonym(s):

1-[8-(2-Chlorophenyl)-9-(4-chlorophenyl)-9H-purin-6-yl]-4-(ethylamino)-4-piperidinecarboxamide hydrochloride, 1-[9-(4-Chlorophenyl)-8-(2-chlorophenyl)-9H-purin-6-yl]-4-ethylaminopiperidine-4-carboxylic acid amide hydrochloride, CP 945598-01, Otenabant hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25Cl2N7O · HCl
CAS Number:
Molecular Weight:
546.88
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

Clc1c(cccc1)c2[n](c4ncnc(c4n2)N5CCC(CC5)(NCC)C(=O)N)c3ccc(cc3)Cl.Cl

InChI

1S/C25H25Cl2N7O.ClH/c1-2-31-25(24(28)35)11-13-33(14-12-25)22-20-23(30-15-29-22)34(17-9-7-16(26)8-10-17)21(32-20)18-5-3-4-6-19(18)27;/h3-10,15,31H,2,11-14H2,1H3,(H2,28,35);1H

InChI key

KPYUQCJBZGQHPL-UHFFFAOYSA-N

Biochem/physiol Actions

CP-945,598 is a potent, selective, high affinity and competitive cannabinoid type 1 (CB1) receptor antagonist.
CP-945,598 is a potent, selective, high affinity and competitive cannabinoid type 1 (CB1) receptor antagonist. CP-945,598 inhibits both basal and cannabinoid agonist-mediated CB1 receptor signaling in vitro and in vivo. CP-945,598 exhibits anorectic activity in two models of acute food intake in rodents, fast-induced re-feeding and spontaneous, nocturnal feeding.
CP-945,598 is also known as 1-(8-(2-Chlorophenyl)-9-(4-chlorophenyl)-9H-purin-6-yl)-4-(ethylamino)piperidine-4-carboxamide. This orally active antagonist of the cannabinoid CB-1 receptor may be used in the treatment of obesity.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Excretion, metabolism, and pharmacokinetics of CP-945,598, a selective cannabinoid receptor antagonist, in rats, mice, and dogs
Miao ZS, et al.
Drug Metabolism and Disposition, 39(12), 2191-2208 (2011)
Lei Xiao et al.
eLife, 7 (2018-04-21)
The experience of rewarding or aversive stimuli is encoded by distinct afferents to dopamine (DA) neurons of the ventral tegmental area (VTA). Several neuromodulatory systems including oxytocin regulate DA neuron excitability and synaptic transmission that process socially meaningful stimuli. We

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service