Skip to Content
MilliporeSigma

Skip To

PCRISPR005

Human CRISPR Inhibition 10x Compatible Druggable Library

Synonym(s):

CRISPR Druggable Library, Human CRISPR Inhibition Kit, Inhibition Library

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 x 200 μL
Please contact Customer Service for Availability
$11,070.00

About This Item

NACRES:
NE.02
UNSPSC Code:
41105904

$11,070.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 5 vials (5x200µL aliquots )

concentration

≥5x108 VP/ml (via p24 Assay)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

General description

The 10x Compatible Human CRISPRi Kinase Phosphatase Drug Targets Kit (Druggable Genome) contains one sub-pool of the top two ranked sgRNAs per gene for increased sensitivity and includes non-targeting controls built-in. In addition, the LV15 vector has both BFP and puromycin as selection markers and an optimized scaffold with capture sequence 2 (CS2, Compatible with 10x Feature Barcode technology) for sgRNA expression and function.

Custom pools for follow-up screening or 10x Genomics Compatible CRISPR pools are also available by contacting your local sales representative.

Application

  • Functional Genomics/Target Validation
  • Focused forward genetic activation screening
  • Validated positive and negative controls
  • Set up and optimization of screen assay
  • Creation of cell lines stably expressing KRAB-dCas9

Biochem/physiol Actions

The power of CRISPR for genome engineering, coupled with the ability to perform large-scale, whole-genome loss-of-function (LOF) screening, has allowed breakthroughs in identifying gene pathways in drug resistance and disease. CRISPR is most commonly used to create double-stranded breaks that often result in loss of gene function (CRISPR-KO). However, the full extent of CRISPR′s utility extends beyond just targeted cutting of DNA. CRISPR′s Nuclease-independent applications provide equal targeting specificity. Instead of cutting, CRISPRi allows for targeted interference of gene function by delivering transcriptional repressor domains to a specific target sequence using modified dCas9 + gRNA complexes. Gene knockdown complements CRISPR-KO and CRISPRa (activation) and has distinct advantages over existing loss-of-function strategies like RNAi. As a world leader in producing lentiviral libraries, we design and build custom pools to any specifications. With this capability, we have partnered with 10x Genomics to bring innovative pooled screening products to enable discovery at single-cell resolution

Features and Benefits

  • Focus on your research, and we will generate your lentivirus screening library.
  • Optimized (F+E) gRNA scaffold. (pools are gRNA-only, KRAB-dCas9 sold separately CRISPRICON or CRISPRI10X
  • A total of 2,318 Human Druggable genes targeted.
  • Total of 4,636 unique gRNAs (top two ranking gRNAs)
  • Use puromycin and or BFP as selection markers

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PCRISPR007HKCRISPRPCRISPR008
application(s)

CRISPR

application(s)

CRISPR

application(s)

CRISPR

application(s)

CRISPR

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

packaging

pkg of 5 vials (5x200µL aliquots )

packaging

pkg of 5 vials (5x200µL aliquots )

packaging

pkg of 8x25 μL (vials)

packaging

pkg of 5 vials (5x200µL aliquots )

concentration

≥5x108 VP/ml (via p24 Assay)

concentration

≥5x108 VP/ml (via p24 Assay)

concentration

5x108  VP/ml (via p24 assay)

concentration

≥5x108 VP/ml (via p24 Assay)

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Could you please confirm if the vector for PCRISPR005, the Human CRISPRi 10x Compatible Druggable Library, is LV15 and not LV15 dual sgRNA?

    1 answer
    1. Yes, that is correct. This is the LV15 backbone, not the dual guide backbone.

      Helpful?

Reviews

No rating value

Active Filters