Skip to Content
Merck

Skip To

P2272

Phosphate buffered saline

liquid, pH 7.2, sterile-filtered, suitable for cell culture

Synonym(s):

Phosphate buffered saline, PBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate buffered saline, pH 7.2, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

Quality Segment

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

pH

7.2

General description

Phosphate-buffered saline (PBS) is a buffer solution used in biological research. It is a water-based salt solution containing sodium phosphate, sodium chloride and, in some formulations, it contains potassium chloride and potassium phosphate. The osmolality and ion concentrations of the solutions match those of the human body (isotonic) and are non-toxic to most cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P4474P5119P4417
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

liquid

form

liquid

form

liquid

form

tablet

pH

7.2

pH

7.4

pH

7.2 (25 °C)

pH

7.2-7.6 (1 tablet/200 mL), 7.4

sterility

sterile-filtered

sterility

sterile-filtered

sterility

autoclaved

sterility

-

impurities

Endotoxin, tested

impurities

Endotoxin, tested

impurities

-

impurities

-


Still not finding the right product?

Explore all of our products under Phosphate buffered saline


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Questions

1–4 of 4 Questions  
  1. I found a publication talking about 0.01 M saline phosphate buffer solution at pH8. I do not understand how to translate 1X PBS solution into molarities. Do you have a clue?

    1 answer
    1. Following is the formulation of the inquired product: 0.01M phosphate buffer, 0.0027 M potassium chloride, and 0.137 M sodium chloride.

      Helpful?

  2. What is the composition of P2272 Phosphate buffered saline pH 7.2, liquid, sterile-filtered, suitable for cell culture ?

    1 answer
    1. The formulation of this Phosphate Buffered Saline product is as follows:
      Potassium Phosphate Monobasic - 210 mg/L
      Sodium Phosphate Dibasic, dihydrate - 482 mg/L
      Sodium Chloride - 9.0 G/L

      Helpful?

  3. Is this calcium-magnesium free?

    1 answer
    1. This PBS formulation does not contain Calcium or Magnesium.

      Helpful?

  4. Is this PBS x1? Or x10? Not specified on the bottle

    1 answer
    1. Helpful?

Reviews

No rating value

Active Filters