Skip to Content
Merck
All Photos(2)

Key Documents

O8264

Sigma-Aldrich

Anti-ORAI1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CRACM1, Anti-Calcium release-activated calcium channel protein 1, Anti-TMEM142A, Anti-Transmembrane protein 142A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~35 kDa (non-glycosylated)
antigen ~50 kDa (glycosylated)

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extract of human HEK-293T cells expressing ORAI1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ORAI1(84876)
mouse ... Orai1(109305)

Related Categories

General description

ORAI calcium release-activated calcium modulator 1 (ORAI1) is the pore-forming subunit of the Ca2+ release-activated (CRAC) channel. The protein possesses four predicted transmembrane domains and intracellular amino and carboxyl-terminal domains. The ORAI1 gene is localized on human chromosome 12q24.31.

Immunogen

synthetic peptide corresponding to amino acids 288-301 of human ORAI1, conjugated to KLH. The corresponding sequence differs by 2 amino acids in rat and mouse.

Application

Anti-ORAI1 antibody produced in rabbit has been used in immunohistochemistry and Western blotting.

Biochem/physiol Actions

ORAI calcium release-activated calcium modulator 1 (ORAI1) modulates Ca2+ entry into cells. The channel is activated by stromal interaction molecule-1 (STIM-1). Mutation in the gene encoding the protein has been linked to late-onset tubular aggregate myopathy and congenital miosis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Store-operated Ca2+ Entry Mediated by Orai1 and TRPC1 Participates to Insulin Secretion in Rat ?-Cells*
Jessica S
The Journal of Biological Chemistry (2015)
Microdeletion of 12q24.31: Report of a Girl With Intellectual Disability, Stereotypies, Seizures and Facial Dysmorphisms
Orazio Palumbo
American Journal of Medical Genetics. Part A (2014)
Yukari Endo et al.
Human molecular genetics, 24(3), 637-648 (2014-09-18)
The store-operated Ca(2+) release-activated Ca(2+) (CRAC) channel is activated by diminished luminal Ca(2+) levels in the endoplasmic reticulum and sarcoplasmic reticulum (SR), and constitutes one of the major Ca(2+) entry pathways in various tissues. Tubular aggregates (TAs) are abnormal structures
Calcium Influx of Mast Cells Is Inhibited by Aptamers Targeting the First Extracellular Domain of Orai1
Renshan Sun
PLoS ONE (2016)
Antibody-Mediated Targeting of the Orai1 Calcium Channel Inhibits T Cell Function
Jennifer H. Cox
PLoS ONE (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service