Skip to Content
Merck
All Photos(1)

Key Documents

N5521

Sigma-Aldrich

α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii)

recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein

Synonym(s):

Acyl-Neuraminyl Hydrolase, Sialidase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

form

buffered aqueous solution

specific activity

≥250 units/mg protein

mol wt

~41 kDa

foreign activity

proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Releases α(2→3)- and α(2→6)-linked N-acetylneuraminic acid from complex oligosaccharides.

Packaging

Provided with 5× reaction buffer (250 mM sodium phosphate, pH 6.0).

Unit Definition

One unit will hydrolyze 1 μmole of 4-methylumbelliferyl α-D-N-acetylneuraminide per min at pH 5.0 at 37 °C

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, and 25 mM NaCl.

Preparation Note

Expressed in glycosidase-free hosts.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodolfo Ocadiz-Delgado et al.
BMC infectious diseases, 13, 20-20 (2013-01-19)
In April 2009, public health surveillance detected an increased number of influenza-like illnesses in Mexico City's hospitals. The etiological agent was subsequently determined to be a spread of a worldwide novel influenza A (H1N1) triple reassortant. The purpose of the
Charles R Beck et al.
Influenza and other respiratory viruses, 7 Suppl 1, 14-24 (2013-02-12)
The objectives of this study were to: (1) reflect on key stages in the discovery, development and pre-pandemic use of neuraminidase inhibitors (NAIs), (2) summarise the evidence of NAI effectiveness for treatment and prophylaxis of seasonal influenza prior to the
Weijia Wang et al.
Journal of virology, 87(8), 4642-4649 (2013-02-15)
In 2009, we successfully produced a high-yield live attenuated H1N1pdm A/California/7/2009 vaccine (CA/09 LAIV) by substitution of three residues (K119E, A186D, and D222G) in the hemagglutinin (HA) protein. Since then, we have generated and evaluated additional H1N1pdm vaccine candidates from
Ramaiah Arunachalam et al.
Interdisciplinary sciences, computational life sciences, 4(4), 282-290 (2013-01-29)
The aim of the present investigation was to discover the genetic relationships of 2009 pandemic novel influenza A/H1N1 virus (NIV) external antigens Hemagglutinin (HA) and Neuraminidase (NA) with other influenza viruses by performing phylogenetic, comparative and statistical analyses. Phylogenetic trees
Kao-Tan Chen et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 37(20), 3068-3073 (2013-01-15)
To isolate and identify active neuraminidase constituents of Polygonum cuspidatum against influenza A (H1N1) influenza virus. On the basis of the bioassay-guided fractionation,such chromatographic methods as silica gel, sephadex LH-20 and HPLC were adopted to isolate active constituents of extracts

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service