Skip to Content
Merck
All Photos(1)

Documents

N0755

Sigma-Aldrich

Netilmicin sulfate salt

Synonym(s):

1-N-Ethylsisomicin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H41N5O7 · 5/2H2SO4
CAS Number:
Molecular Weight:
720.78
EC Number:
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.85

form

powder

Quality Level

color

white to faint yellow

solubility

H2O: 10 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

InChI

1S/2C21H41N5O7.5H2O4S/c2*1-4-26-13-7-12(24)16(32-19-11(23)6-5-10(8-22)31-19)14(27)17(13)33-20-15(28)18(25-3)21(2,29)9-30-20;5*1-5(2,3)4/h2*5,11-20,25-29H,4,6-9,22-24H2,1-3H3;5*(H2,1,2,3,4)/t2*11-,12+,13-,14+,15-,16-,17+,18-,19-,20-,21+;;;;;/m11...../s1

InChI key

AGFWIZQEWFGATK-UNZHCMSXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Netilmicin is an aminoglycoside antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studies and in susceptibility studies of Pseudomonas aeruginosa and Pseudomonas pseudomallei .

Biochem/physiol Actions

An aminoglycoside antibiotic active against most Gram-negative and some Gram-positive bacteria, including many gentamycin-resistant strains. The antibiotic enters the cell through a porin channel and binds to the 30S ribosomal subunit, causing misreading of mRNA.
Netilmicin irreversibly binds to specific 30S-subunit proteins and 16S rRNA. Netilmicin binds to four nucleotides of 16S rRNA and a single amino acid of protein S12. This interferes with the decoding site in the vicinity of nucleotide 1400 in 16S rRNA of 30S subunit. This region interacts with the wobble base in the anticodon of tRNA. This leads to interference with the initiation complex, misreading of mRNA so incorrect amino acids are inserted into the polypeptide. This results in nonfunctional and toxic peptides and the bacterium is unable to synthesize crucial proteins.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Fu You et al.
Antimicrobial agents and chemotherapy, 53(10), 4525-4528 (2009-07-29)
Vertilmicin is a novel aminoglycoside antibiotic with potent activity against gram-negative and -positive bacteria in vitro. In this study, we further evaluated the efficacy of vertilmicin in vivo in systemic and local infection animal models. We demonstrated that vertilmicin had
Arul Joseph et al.
Journal of chromatographic science, 48(7), 607-612 (2010-09-08)
Netilmicin is a semi-synthetic aminoglycoside antibiotic used against a broad spectrum of Gram-negative bacteria. A reversed-phase high-performance liquid chromatographic method has been developed to determine the composition of netilmicin sulfate and to estimate its related substances without pre- or post-column
Philip K N Lam et al.
Antimicrobial agents and chemotherapy, 54(2), 963-965 (2009-12-02)
Adsorption of gentamicin and netilmicin by new polyacrylonitrile and polyamide hemofiltration filters was studied over 4 h, using a single-compartment in vitro continuous venovenous hemofiltration model. After the first dose (16.6 mg of gentamicin, 19.3 mg of netilmicin), 14.9 mg
D W McEniry et al.
The Journal of antimicrobial chemotherapy, 21(2), 171-175 (1988-02-01)
Pseudomonas pseudomallei is resistant to many antimicrobial agents. We determined the susceptibility to twelve beta-lactams and six aminoglycosides of twelve isolates of P. pseudomallei. Imipenem (MIC range 0.5-1.0 mg/l; MIC90 1.0 mg/l), ceftazidime (MIC range 1-8 mg/l; MIC90 8.0 mg/l)
"Typhlitis" in rectum.
Unsal Ozgen et al.
Pediatrics international : official journal of the Japan Pediatric Society, 52(1), e32-e33 (2010-02-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service