Skip to Content
MilliporeSigma

MTCC200050

MTC Bio SureSeal S Microcentrifuge Tube

capacity 1.5 mL, sterile, clear, pack of 500 ea (10 x packs 50ea)

Synonym(s):

centrifuge tube, plastic tube, MTC Bio C2000-50, sample collection tube, sample receptacle, sample tube

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41121703
Capacity:
1.5 mL
Material:
clear , polypropylene , round
Sterility:
sterile
Packaging:
pack of 500 ea (10 x packs 50ea)
Parameter:
28,000 × g speed
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , polypropylene , round

sterility

sterile

packaging

pack of 500 ea (10 x packs 50ea)

manufacturer/tradename

MTC Bio C2000-50

parameter

28,000 × g speed

capacity

1.5 mL

General description

Designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation, and general sample/reagent storage. SureSeal(TM) S tubes are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity, centrifugal strength (tested up to 28,000×g) and the usable temperature range (-90°C to +121°C). The full-sized tube cap (with frosted marking surface), combined with the unique ring and groove closure, enhances the security of the liquid-tight seal, while minimizing the required locking forces for a more ergonomic opening/closing of the tube caps.
Microtube w/ cap, 1.5ml, clear, sterile, w/ self-standing bag, 10 packs of 50, 500/pk

Features and Benefits

  • Sterile comes standard – by electron beam radiation
  • Self-Standing bag – with gusseted bottom
  • Stop-Pops supplied – 10 cap locks in every bag

Legal Information

MTC is a trademark of MTC Bio
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Stop-Pops is a trademark of MTC Bio
SureSeal S is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library