Skip to Content
MilliporeSigma

Skip To

M8902

MES hemisodium salt

≥98% (titration)

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid hemisodium salt, 4-Morpholineethanesulfonic acid hemisodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
$66.10
100 g
Check Cart for Availability
$197.00
$167.45

About This Item

Empirical Formula (Hill Notation):
C6H13NO4S·0.5Na
CAS Number:
Molecular Weight:
206.73
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25
MDL number:

$66.10


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (titration)

form

crystalline powder

impurities

≤8% water content (Karl Fischer)

useful pH range

5.5-6.7

pKa 

6.1

solubility

water: 500 mg/mL, clear, colorless

cation traces

Pb: ≤5 ppm

SMILES string

[Na+].OS(=O)(=O)CCN1CCOCC1.[O-]S(=O)(=O)CCN2CCOCC2

InChI

1S/2C6H13NO4S.Na/c2*8-12(9,10)6-3-7-1-4-11-5-2-7;/h2*1-6H2,(H,8,9,10);/q;;+1/p-1

InChI key

KHQSUKMBWPKXOP-UHFFFAOYSA-M

Application

MES hemisodium salt has been used as a buffer for the resuspension of carboxylated micelles.[1][2] It has also been used as a buffer for the immobilization of antibacterial nisin peptides.

Biochem/physiol Actions

MES is one of the major Good′s buffers and is applicable in regulating the pH value in physiological experiments, reagent solutions and plant culture medium.[3]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M9027M0164M3885
assay

≥98% (titration)

assay

≥99% (titration)

assay

-

assay

≥99% (titration)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

400

form

crystalline powder

form

powder or crystals

form

dry powder

form

crystalline powder

solubility

water: 500 mg/mL, clear, colorless

solubility

water: 500 mg/mL, clear, colorless to almost colorless

solubility

-

solubility

water: 0.5 g/mL, clear, colorless

cation traces

Pb: ≤5 ppm

cation traces

-

cation traces

-

cation traces

-

impurities

≤8% water content (Karl Fischer)

impurities

-

impurities

-

impurities

≤8% water content (Karl Fischer)


Still not finding the right product?

Explore all of our products under MES hemisodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters