Skip to Content
Merck
All Photos(1)

Key Documents

LUC1

Sigma-Aldrich

Luciferase Reporter Gene Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.84

usage

 kit sufficient for 100 assays

storage temp.

−70°C

General description

Firefly luciferase is one of the most utilized reporter genes for the study of gene expression. It is an extremely sensitive, rapid, and easy-to-use reporter gene. The chemiluminescent reaction catalyzed by luciferase is one of the most sensitive analytical tools for measuring gene expression. The luciferase substrate contains coenzyme A for increased and sustained luminescence compared to conventional methods. This eliminates the need for automated luminometer injection of the substrate and allows analysis by photographic film or scintillation counting. The lysis buffer contains polymyxin B which further enhances the signal and eliminates lysozyme treatment and freeze-thawing of cells. Cell lysis buffer is compatible with β-galactosidase assays. The enzyme encoded by the luciferase reporter gene catalyzes the oxidation of D-luciferin in the presence of ATP, oxygen, and Mg2+. The fluorescent product is quantified to measure its activity.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerben van Ooijen et al.
Current biology : CB, 21(10), 869-875 (2011-05-03)
Circadian clocks were, until recently, seen as a consequence of rhythmic transcription of clock components, directed by transcriptional/translational feedback loops (TTFLs). Oscillations of protein modification were then discovered in cyanobacteria. Canonical posttranslational signaling processes have known importance for clocks across
Paola Monti et al.
Scientific reports, 10(1), 18427-18427 (2020-10-30)
Chronic lymphocytic leukaemia (CLL) is characterised by a heterogeneous clinical course. Such heterogeneity is associated with a number of markers, including TP53 gene inactivation. While TP53 gene alterations determine resistance to chemotherapy, it is not clear whether they can influence
Jaya Mishra et al.
Journal of the American Society of Nephrology : JASN, 14(10), 2534-2543 (2003-09-30)
Acute renal failure (ARF) secondary to ischemic injury remains a common and potentially devastating problem. A transcriptome-wide interrogation strategy was used to identify renal genes that are induced very early after renal ischemia, whose protein products might serve as novel
Stephen T Smale
Cold Spring Harbor protocols, 2010(5), pdb-pdb (2010-05-05)
When a transient or stable transfection assay is developed for a promoter, a primary objective is to quantify promoter strength. Because transfection efficiency in such assays can be low, promoters are commonly fused to heterologous reporter genes that encode enzymes
Giulia Donvito et al.
Neuropharmacology, 148, 320-331 (2018-03-24)
Cigarette smokers with brain damage involving the insular cortex display cessation of tobacco smoking, suggesting that this region may contribute to nicotine addiction. In the present study, we speculated that molecules in the insular cortex that are sensitive to experimental

Articles

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service