Skip to Content
Merck
All Photos(1)

Key Documents

L8402

Sigma-Aldrich

Lysozyme from human neutrophils

≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)

Synonym(s):

Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

human neutrophils

Quality Level

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

specific activity

≥100,000 units/mg protein (E1%/280)

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

SMILES string

N1([C@@H](CCC1)C(=O)N[C@@H](CCC(=O)N)C(=O)NCC(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](CCCN\C(=N\[H])\N)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc4c5c([nH]c4)cccc5)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc2c3c([nH]c2)cccc3)C(=O)N[C@@H](CCCN\C(=N\[H])\N)C(=O)N[C@@H]

InChI

1S/C99H159N37O23/c1-11-50(8)77(132-72(139)46-120-81(145)63(32-33-70(101)137)124-88(152)69-31-21-39-136(69)93(157)68(43-73(140)141)131-84(148)62(29-19-37-116-98(109)110)125-90(154)75(48(4)5)135-91(155)76(49(6)7)133-80(144)57(100)24-16-34-113-95(103)104)92(156)126-60(27-17-35-114-96(105)106)82(146)121-51(9)78(142)129-66(41-54-45-119-59-26-15-13-23-56(54)59)87(151)134-74(47(2)3)89(153)122-52(10)79(143)128-65(40-53-44-118-58-25-14-12-22-55(53)58)85(149)123-61(28-18-36-115-97(107)108)83(147)130-67(42-71(102)138)86(150)127-64(94(158)159)30-20-38-117-99(111)112/h12-15,22-23,25-26,44-45,47-52,57,60-69,74-77,118-119H,11,16-21,24,27-43,46,100H2,1-10H3,(H2,101,137)(H2,102,138)(H,120,145)(H,121,146)(H,122,153)(H,123,149)(H,124,152)(H,125,154)(H,126,156)(H,127,150)(H,128,143)(H,129,142)(H,130,147)(H,131,148)(H,132,139)(H,133,144)(H,134,151)(H,135,155)(H,140,141)(H,158,159)(H4,103,104,113)(H4,105,106,114)(H4,107,108,115)(H4,109,110,116)(H4,111,112,117)/t50-,51-,52-,57-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,74-,75-,76-,77-/m0/s1

InChI key

ZJCXKXFAOCLSRV-RYLVIXIQSA-N

Gene Information

human ... LYZ(4069)

Looking for similar products? Visit Product Comparison Guide

General description

Lysozyme is abundantly found in animal and plant kingdoms. It is a natural food preservative and also found to be present in specific bacterial cell walls. Lysozyme is usually present in tears, milk, urine and saliva.

Application

Lysozyme from human neutrophils has been used in the checkerboard method for establishing a synergy between peptidomimetic 1 and immune effector molecules against E. coli.

Biochem/physiol Actions

Lysozymes participates in the defense mechanism. It has the ability to stimulate catalysis by bringing steric stress in the substrates.

Unit Definition

One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.

Physical form

Lyophilized from 50 mM sodium acetate, pH 6.0, with 100 mM NaCl

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selectivity in the potentiation of antibacterial activity of alpha-peptide beta-peptoid peptidomimetics and antimicrobial peptides by human blood plasma
Hein-Kristensen L, et al.
Research in Microbiology, 164(9), 933-940 (2013)
María Victoria Humbert et al.
PLoS pathogens, 13(6), e1006448-e1006448 (2017-07-01)
Pathogenic and commensal Neisseria species produce an Adhesin Complex Protein, which was first characterised in Neisseria meningitidis (Nm) as a novel surface-exposed adhesin with vaccine potential. In the current study, the crystal structure of a recombinant (r)Nm-ACP Type I protein
Maj Hedtjärn et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 24(12), 1333-1351 (2004-12-31)
Brain ischemia triggers an inflammatory reaction that progresses for days to weeks and seems to have a role in secondary progression of injury. Inflammation induces a complex pattern of signaling molecules with partly contradictory actions, and the responses may be
Natural Food Antimicrobial Systems (2000)
Masaki Honda et al.
Transplantation, 95(4), 551-558 (2013-02-21)
Neutrophils are considered responsible for the pathophysiologic changes during hepatic ischemia-reperfusion (I/R) injury; however, few studies have examined real-time intravital neutrophil recruitment. Here, we show a method for imaging the neutrophil recruitment in hepatic I/R injury using two-photon laser scanning

Protocols

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

This enzymatic rate determination may be used for Lysozyme products. It is not to be used to assay recombinant or insoluble Lysozyme on agarose.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service