Skip to Content
Merck
All Photos(1)

Documents

L7035

Sigma-Aldrich

clasto-Lactacystin β-lactone

Synonym(s):

(1R,4R,5S)-1-[(1S)-1-Hydroxy-2-Methylpropyl]-4-Methyl-6-Oxa-2-Azabicyclo[3.2.0]Heptane-3,7-Dione, Omuralide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO4
CAS Number:
Molecular Weight:
213.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

film

solubility

DMSO: soluble 25 mg/mL

storage temp.

−20°C

SMILES string

[H][C@@]12OC(=O)[C@@]1(NC(=O)[C@@H]2C)[C@@H](O)C(C)C

InChI

1S/C10H15NO4/c1-4(2)6(12)10-7(15-9(10)14)5(3)8(13)11-10/h4-7,12H,1-3H3,(H,11,13)/t5-,6+,7+,10-/m1/s1

InChI key

FWPWHHUJACGNMZ-NBBQQVJHSA-N

Application

Clasto-Lactacystin β-lactone has been used as a proteasomal inhibitor.
Clasto-Lactacystin β-lactone has been used as a protein degradation inhibitor to test its effect on memory improvements in mice. It has also been used as a proteasome inhibitor in human ovarian surface epithelium (HOSE) cells and dendritic cells (DCs).

Biochem/physiol Actions

Cell-permeable and irreversible proteasome inhibitor. Lactacystin acts as a precursor for clasto-lactacystin β-lactone.
Clasto-Lactacystin β-lactone (cLβL) is synthesized from lactacystin. It is cell-permeable and cLβL acts on the N-terminal threonine of subunit proteasome β -subunit X It also inhibits 20S proteasome activity in Haloferax volcanii by acting in the N-threonine residue of the  β -type subunits.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristen Willeumier et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 26(44), 11333-11341 (2006-11-03)
The ubiquitin proteasome system, generally known for its function in protein degradation, also appears to play an important role in regulating membrane trafficking. A role for the proteasome in regulating presynaptic release and vesicle trafficking has been proposed for invertebrates
Coordinated regulation of dendrite arborization by epigenetic factors CDYL and EZH2
Qi C, et al.
The Journal of Neuroscience, 34(13), 4494-4508 (2014)
Florian Vogel et al.
Traffic (Copenhagen, Denmark), 8(9), 1205-1214 (2007-07-17)
Intracellular trafficking of viral movement proteins (MPs) in plants has mainly been studied using Tobacco mosaic virus MP30 (TMV MP30) as a model system. Because of the limitations of TMV MP30 expression in Arabidopsis thaliana, these studies have mostly been
Sue-Hyun Lee et al.
Science (New York, N.Y.), 319(5867), 1253-1256 (2008-02-09)
Reactivated memory undergoes a rebuilding process that depends on de novo protein synthesis. This suggests that retrieval is dynamic and serves to incorporate new information into preexisting memories. However, little is known about whether or not protein degradation is involved
A K Ho et al.
Endocrinology, 148(2), 743-751 (2006-11-04)
In this study, we investigated the role of two inducible repressor proteins, inducible cAMP early repressor (ICER) and Fos-related antigen 2 (Fra-2) in the adrenergic induction of MAPK phosphatase-1 (MKP-1) as compared with their roles in the induction of arylalkylamine-N-acetyltransferase

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service