Skip to Content
Merck
All Photos(3)

Key Documents

L5768

Sigma-Aldrich

Lanosterol

≥93%, powder

Synonym(s):

3β-Hydroxy-8,24-lanostadiene, 8,24-Lanostadien-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥93%

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

[H][C@@]1(CC[C@@]2(C)C3=C(CC[C@]12C)[C@@]4(C)CC[C@H](O)C(C)(C)[C@]4([H])CC3)[C@H](C)CC\C=C(/C)C

InChI

1S/C30H50O/c1-20(2)10-9-11-21(3)22-14-18-30(8)24-12-13-25-27(4,5)26(31)16-17-28(25,6)23(24)15-19-29(22,30)7/h10,21-22,25-26,31H,9,11-19H2,1-8H3/t21-,22-,25+,26+,28-,29-,30+/m1/s1

InChI key

CAHGCLMLTWQZNJ-BQNIITSRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lanosterol, an amphipathic molecule, that is produced by lanosterol synthase (LSS). It is enriched in the lens.

Application

Lanosterol has been used:
  • as a standard in HPLC for the quantification in testis samples
  • in S-adenosyl-L-methionine:Δ24-sterol-C-methyltransferase (SMT) assay
  • to treat wild-type cells growing in rich medium to know its effects on Sre1 protein

Biochem/physiol Actions

Cholesterol precursor sterol.
Lanosterol serves as an endogenous selective modulator of macrophage immunity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorena Fernández-Cabezón et al.
Frontiers in microbiology, 9, 958-958 (2018-06-06)
Nowadays steroid manufacturing occupies a prominent place in the pharmaceutical industry with an annual global market over $10 billion. The synthesis of steroidal active pharmaceutical ingredients (APIs) such as sex hormones (estrogens, androgens, and progestogens) and corticosteroids is currently performed
Identification and Characterization of an S-Adenosyl-L-methionine:-Sterol-C-methyltransferase cDNA from Soybean
Shi J, et al.
The Journal of Biological Chemistry, 271(16), 9384-9389 (1996)
Lanosterol reverses protein aggregation in cataracts
Zhao L, et al.
Nature, 523(7562), 607-607 (2015)
Marja-Riitta Taskinen et al.
Journal of clinical lipidology, 14(1), 77-87 (2020-01-10)
Monoclonal antibodies to proprotein convertase subtilisin/kexin type 9 (PCSK9) significantly lower the levels of low-density lipoprotein and very-low-density lipoproteins (VLDL), but their effect on postprandial lipoprotein metabolism in dyslipidemic subjects is unclear. This study aimed to investigate the effects of
Ying-Chi Du et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(8-9), 788-796 (2012-04-21)
Antrodia camphorata (AC) is a native Taiwanese mushroom which is used in Asian folk medicine as a chemopreventive agent. The triterpenoid-rich fraction (FEA) was obtained from the ethanolic extract of AC and characterized by high performance liquid chromatography (HPLC). FEA

Articles

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service