Skip to Content
Merck
All Photos(1)

Key Documents

I7125

Sigma-Aldrich

5-Iodo-2′-deoxyuridine

≥99% (HPLC)

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)-5-iodouracil, 2′-Deoxy-5-iodouridine, 5-IUdR, IDU, IdUrd, Idoxuridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11IN2O5
CAS Number:
Molecular Weight:
354.10
Beilstein:
30397
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥99% (HPLC)

form

powder or crystals

mp

194 °C (lit.)

solubility

1 N NH4OH: 50 mg/mL, clear to very slightly hazy, colorless to light yellow

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(I)C(=O)NC2=O

InChI

1S/C9H11IN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

XQFRJNBWHJMXHO-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Iodo-2′-deoxyuridine (5-IUdR; IdUrd) is a pyrimidine analog (halogenated thymidine) that has antiviral activity agains vaccinia virus (orthopoxviruses). IdUrd is used as a radiosensitizer in human cancer studies.
5-Iodo-2′-deoxyuridine has been used in molecular combing assay or DNA fiber assay.

Biochem/physiol Actions

5-Iodo-2′-deoxyuridine prevents in vitro DNA viral replication. This is observed in herpesviruses and poxviruses It might possess teratogenic, tumor-promoting, mutagenic, and immunosuppressive properties. 5-Iodo-2′-deoxyuridine, used in topical applications, is effective against epithelial infections.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antivirals against Herpes Viruses
Mandell, Douglas, and Bennett's Principles and Practice of Infectious Diseases, 546-562 (2015)
Johan Neyts et al.
Antimicrobial agents and chemotherapy, 46(9), 2842-2847 (2002-08-17)
There is a concern that there may be unregistered stocks of smallpox that can be used for bioterrorism or biological warfare. According to the WHO Advisory Committee on Variola Research, there is a need to develop strategies to treat smallpox
Jan Kajstura et al.
Circulation research, 107(2), 305-315 (2010-06-05)
The ability of the human heart to regenerate large quantities of myocytes remains controversial, and the extent of myocyte renewal claimed by different laboratories varies from none to nearly 20% per year. To address this issue, we examined the percentage
Qiuwei Pan et al.
Gastroenterology, 144(4), 761-770 (2012-12-26)
The existence of slowly cycling, adult stem cells has been challenged by the identification of actively cycling cells. We investigated the existence of uncommitted, slowly cycling cells by tracking 5-iodo-2'-deoxyuridine (IdU) label-retaining cells (LRCs) in normal esophagus, Barrett's esophagus (BE)
Esther G C Troost et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 51(5), 713-719 (2010-04-17)
Accelerated tumor cell repopulation is an important mechanism adversely affecting therapeutic outcome in head and neck cancer. The noninvasive assessment of the proliferative state of a tumor by PET may provide a selection tool for customized treatment. 3'-deoxy-3'-(18)F-fluorothymidine ((18)F-FLT) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service