Skip to Content
Merck
All Photos(2)

Key Documents

I6035

Sigma-Aldrich

L-Iditol

≥98% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (GC)

form

solid

color

white

mp

78-80 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

OC[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5+,6+/m0/s1

InChI key

FBPFZTCFMRRESA-UNTFVMJOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A rare sugar alcohol (polyol).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N A Williams et al.
Journal of parenteral science and technology : a publication of the Parenteral Drug Association, 47(3), 119-123 (1993-05-01)
The mechanical properties of frozen mannitol, L-iditol, dulcitol, and sorbitol solutions were measured as a function of temperature during warming (after freezing) using a thermal mechanical analyzer (TMA). The mannitol sample first underwent a contractive phase starting at 30 degrees
Constitution of the dianhydrides of sorbitol and iditol.
R MONTGOMERY et al.
Nature, 157, 372-372 (1946-03-23)
Wojciech Schönemann et al.
Bioorganic & medicinal chemistry, 18(7), 2645-2650 (2010-03-17)
A short synthesis of new beta-1-C-alkyl-1,5-dideoxy-1,5-imino-l-iditols by means of the diastereoselective addition of Grignard reagents onto a glucopyranosylamine is described. These compounds were evaluated as beta-glucocerebrosidase inhibitors and their activity was compared with that of related iminosugar derivatives in the
Kazumasa Oosaka
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 129(5), 575-580 (2009-05-08)
Allitol, D-talitol and L-iditol are sugar alcohols that are rare in nature. Due to their previous rarity, little is known about the laxative effects of these rare sugar alcohols. Therefore, reliable data on the laxative effect that these sugar alcohols
C A Brearley et al.
The Biochemical journal, 314 ( Pt 1), 215-225 (1996-02-15)
We have undertaken an analysis of the inositol phosphates of Spirodela polyrhiza at a developmental stage when massive accumulation of InsP6 indicates that a large net synthesis is occurring. We have identified Ins3P, Ins(1,4)P2, Ins(3,4)P2 and possibly Ins(4,6)P2, Ins(3,4,6)P3, Ins(3,4,5,6)P4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service