Skip to Content
Merck
All Photos(5)

Key Documents

I0879

Sigma-Aldrich

Inosine 5′-triphosphate trisodium salt

≥95%

Synonym(s):

ITP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12Na3N4O14P3
CAS Number:
Molecular Weight:
574.11
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥95%

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na].OC1C(O)C(OC1COP(O)(=O)OP(O)(=O)OP(O)(O)=O)n2cnc3C(=O)N=CNc23

InChI

1S/C10H15N4O14P3.Na.H/c15-6-4(1-25-30(21,22)28-31(23,24)27-29(18,19)20)26-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;/h2-4,6-7,10,15-16H,1H2,(H,21,22)(H,23,24)(H,11,12,17)(H2,18,19,20);;

InChI key

RIERCUPIZLBNFQ-UHFFFAOYSA-N

Gene Information

human ... HRAS(3265)

Looking for similar products? Visit Product Comparison Guide

Application

Inosine 5′-triphosphate (ITP) is used in studies on the impact of deamination of ATP and GTP by various enzymes and chemical processes. ITP may be used as a substrate to study the specificity and kinetics of nucleoside-5′-triphosphatase and ATPase. ITP is also used to study activation of various ATPases and GTPases.
Inosine 5′-triphosphate trisodium salt has been used to investigate its influence on the chromosome aberration rate, the mitotic rate, sister-chromatid exchange (SCE) frequency, the proportion of first (X1), second (X2) and third (X3) division metaphases of human peripheral lymphocytes.

Biochem/physiol Actions

Inosine 5′-triphosphate (ITP) has the ability to support the initiation of effector system. It can prevent guanosine 5′-triphosphate (GTP) hydrolysis, that is catalysed by transducin (TD). ITP can proficiently induce secretion in permeabilized cells than GTP. Instead of GTP, ITP can be used for the initiation and the elongation steps of reovirus transcription. ITP can also replace GTP in the activation of G-protein.

Preparation Note

Prepared from Bacterial ATP

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

As to the clastogenic-, sister-chromatid exchange inducing-and cytotoxic activity of inosine triphosphate in cultures of human peripheral lymphocytes
Vormittag W and Brannath W
Mutation Research, 476(1), 71-81 (2001)
T Toyo-oka
Circulation research, 49(6), 1350-1355 (1981-12-01)
The contraction of cardiac muscle that has been treated with glycerol requires Ca2+ (pCa 8-5), when MgATP is used as a substrate. In contrast, this preparation contracts, even in the absence of Ca2+ (pCa 8-10), when ATP is replaced by
D Pollard-Knight et al.
The Biochemical journal, 245(3), 625-629 (1987-08-01)
When ATP, the normal phosphate donor for hexokinase D ('glucokinase'), is replaced by ITP, the positive co-operativity with respect to glucose disappears. This may be rationalized in relation to kinetic models for hexokinase D co-operativity, which assume that with the
Adenosine cyclic 3',5'-monophosphate dependent protein kinase: kinetic mechanism for the bovine skeletal muscle catalytic subunit.
P F Cook et al.
Biochemistry, 21(23), 5794-5799 (1982-11-09)
Initiation of reovirus transcription by inosine 5'-triphosphate and properties of 7-methylinosine-capped, inosine-substituted messenger ribonucleic acids
Morgan M A and Shatkin A J
Biochemistry, 19(26), 5960-5966 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service