Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
description
For use in the Silver Stain Kit No. HT100-A
Quality Segment
form
solution
shelf life
Expiry date on the label
IVD
for in vitro diagnostic use
concentration
2 g/dL in deionized water
application(s)
hematology
histology
storage temp.
2-8°C
SMILES string
[Na+].[Na+].[O-]S([O-])(=O)=S
InChI
1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key
AKHNMLFCWUSKQB-UHFFFAOYSA-L
Application
1 of 1
This Item | |||
|---|---|---|---|
| form solution | form powder | form powder, crystals or chunks | form crystals |
| application(s) hematology | application(s) - | application(s) - | application(s) - |
| storage temp. 2-8°C | storage temp. - | storage temp. - | storage temp. - |
| description For use in the Silver Stain Kit No. HT100-A | description - | description - | description - |
| Quality Level 500 | Quality Level - | Quality Level 200 | Quality Level 200 |
| shelf life Expiry date on the label | shelf life - | shelf life - | shelf life - |
Still not finding the right product?
Explore all of our products under Sodium Thiosulfate Solution
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| HT1005-100ML | 04061838207388 |
