Key Documents
HS24490M
Lab Alert® triple timer and clock
About This Item
UNSPSC Code:
41122411
NACRES:
NB.51
manufacturer/tradename
Heathrow Scientific HS24490M
Features and Benefits
  • Three channels to count up/down with
  • Simultaneous display, time-out, and 60 second audible alarm
  • Channels may be started independently or simultaneously
  • Timer automatically counts
  • 86 x 66 x 22 mm
Legal Information
Lab Alert is a registered trademark of Heathrow Scientific