Skip to Content
Merck
All Photos(10)

Key Documents

HPA029524

Sigma-Aldrich

Anti-SEPT7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-BRICD4, Anti-CDC10, Anti-CDC3, Anti-ChM1L, Anti-SEPT7A, Anti-TEM, Anti-myodulin, Anti-septin 7, Anti-tendin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

VNIIPLIAKADTLTPEECQQFKKQIMKEIQEHKIKIYEFPETDDEEENKLVKKIKDRLPLAVVGSNTIIEVNGKRVRGRQYPW

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEPTIN7(989)

General description

SEPT7 (Septin 7) is a 418 amino acid protein and contains a guanosine triphosphate (GTP)-binding domain. The gene is mapped to human chromosome 7p14. It is a member of the polymerizing GTPases family.

Immunogen

septin 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SEPT7 antibody produced in rabbit has been used in western blotting and immunofluorescence.

Biochem/physiol Actions

Septins (SEPTs) are important proteins which regulate microtubule and actin dynamics. SEPT7 is associated with actin assembly and cell morphology. SEPT7 forms a complex with SEPT2 and SEPT6. In addition, it binds to CENP-E (centromere protein E). The association is needed for the localization of CENP-E to the kinetochore and for correct chromosomal alignment. In endothelial cells, it participates in the internalization of Candida albicans.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74944

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Septins regulate actin organization and cell-cycle arrest through nuclear accumulation of NCK mediated by SOCS7.
Kremer BE, et al.
Cell, 130, 837-850 (2007)
Septins 2, 7 and 9 and MAP4 colocalize along the axoneme in the primary cilium and control ciliary length.
Ghossoub R, et al.
Journal of Cell Science, 126, 2583-2594 (2013)
Septin 7 interacts with centromere-associated protein E and is required for its kinetochore localization.
Zhu M, et al.
The Journal of Biological Chemistry, 283, 18916-18925 (2008)
Role of endothelial cell septin 7 in the endocytosis of Candida albicans.
Phan QT, et al.
mBio, 4, e00542-e00513 (2013)
MiR-30a-5p antisense oligonucleotide suppresses glioma cell growth by targeting SEPT7.
Jia Z, et al.
PLoS ONE, 8, e55008-e55008 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service