Skip to Content
Merck
All Photos(2)

Key Documents

HPA016909

Sigma-Aldrich

Anti-DCBLD2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, Anti-Discoidin, CUB and LCCL domain-containing protein 2, Anti-Endothelial and smooth muscle cell-derived neuropilin-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AWHWRNRKKKTEGTYDLPYWDRAGWWKGMKQFLPAKAVDHEETPVRYSSSEVNHLSPREVTTVLQADSAEYAQPLVGGIVGTLHQRSTFKPEEGKEAGYADLDPYNSPGQEVYHAYAEPLPITGPEYATPIIMDMSGHPTTSVGQPST

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCBLD2(131566)

General description

DCBLD2 (discoidin, CUB and LCCL domain containing 2) is a novel neuropilin-like type-I transmembrane protein belonging to the neuropilin family. It is expressed in endothelial cells specifically in vascular smooth muscle cells. It is composed of a CUB domain and a coagulation factor V/VIII homology domain.

Immunogen

Discoidin, CUB and LCCL domain-containing protein 2 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-DCBLD2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

DCBLD2 (discoidin, CUB and LCCL domain containing 2) is highly involved in the regulation of vascular cell growth. It possesses tumor suppressive property, where it can suppress the cell migration signaling, and invasion of cancer cells. It acts as a potential gene for the miR-451a-mediated regulation in hypopharyngeal squamous cell carcinoma (HSCC). DCBLD2 regulates PDGFRβ (Platelet-derived growth factor receptor β) signaling by modulating receptor ubiquitination in vascular smooth muscle cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71630

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pichai Raman et al.
PloS one, 13(8), e0201751-e0201751 (2018-08-10)
Pancreatic ductal adenocarcinoma (PDAC) is the third leading cause of cancer death in the US. Despite multiple large-scale genetic sequencing studies, identification of predictors of patient survival remains challenging. We performed a comprehensive assessment and integrative analysis of large-scale gene
I Fukumoto et al.
British journal of cancer, 111(2), 386-394 (2014-06-12)
Hypopharyngeal squamous cell carcinoma (HSCC) has a very poor prognosis because of its high rates of regional and distant metastasis. Identification of differentially expressed miRNAs and their regulated molecular targets in tumour cells might enhance our understanding of the molecular
K Kobuke et al.
The Journal of biological chemistry, 276(36), 34105-34114 (2001-07-12)
A novel cDNA has been isolated from primary culture of human coronary arterial cells by a signal sequence trap method, and designated ESDN (endothelial and smooth muscle cell-derived neuropilin-like molecule). ESDN is a type-I transmembrane protein with the longest cleavable
Xiaojia Guo et al.
The Journal of biological chemistry, 284(43), 29376-29382 (2009-08-22)
Endothelial and smooth muscle cell-derived neuropilin-like protein (ESDN) is up-regulated in the neointima of remodeling arteries and modulates vascular smooth muscle cell (VSMC) growth. Platelet-derived growth factor (PDGF) is the prototypic growth factor for VSMCs and plays a key role
Lei Nie et al.
The Journal of clinical investigation, 123(12), 5082-5097 (2013-11-02)
Aberrant blood vessel formation contributes to a wide variety of pathologies, and factors that regulate angiogenesis are attractive therapeutic targets. Endothelial and smooth muscle cell-derived neuropilin-like protein (ESDN) is a neuropilin-related transmembrane protein expressed in ECs; however, its potential effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service