Skip to Content
Merck
All Photos(1)

Key Documents

H8885

Sigma-Aldrich

Hydroxyzine dihydrochloride

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27ClN2O2 · 2HCl
CAS Number:
Molecular Weight:
447.83
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97%

form

powder

solubility

H2O: soluble
alcohol: soluble
chloroform: soluble

SMILES string

Cl[H].Cl[H].OCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3

InChI

1S/C21H27ClN2O2.2ClH/c22-20-8-6-19(7-9-20)21(18-4-2-1-3-5-18)24-12-10-23(11-13-24)14-16-26-17-15-25;;/h1-9,21,25H,10-17H2;2*1H

InChI key

ANOMHKZSQFYSBR-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxyzine dihydrochloride (HDH) is the first-generation antihistamine, that belongs to the piperazine class, an H1 receptor antagonist.

Application

Hydroxyzine dihydrochloride has been used in the study of comparative anxiolytic activity of various drugs in experimental models of rats.

Biochem/physiol Actions

Hydroxyzine dihydrochloride (HDH) is used to regulate anxiety. It is also used to treat bronchial asthma and to relax patients before surgery.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Acid-base titrimetric assay of hydroxyzine dihydrochloride in pharmaceutical samples
Rajendraprasad N, et al.
Chemical Industry Times / Hua Gong Shi Kan, 16(2), 127-132 (2010)
COMPARATIVE ANXIOLYTIC ACTIVITY OF VARIOUS DRUGS IN EXPERIMENTAL MODELS OF RATS
Vitthal K R, et al.
International Journal of Current Research and Review, 4(24), 77-77 (2012)
Sunita B Chhaiya et al.
Indian journal of dermatology, venereology and leprology, 78(5), 605-610 (2012-09-11)
Ivermectin has opened a new era in the management of scabies as orally effective drug. However, topical route has been little explored for ivermectin. To compare the efficacy and safety of topical permethrin, oral ivermectin, and topical ivermectin in the
Cristina Lopez-Peig et al.
BMC research notes, 5, 684-684 (2012-12-15)
Benzodiazepine (BZD), the long-term treatment of which is harmful for cognitive function, is widely prescribed by General Practitioners in Spain. Based on studies performed in other countries we designed a nurse-led BZD withdrawal program adapted to Spanish Primary Care working
N Javaud et al.
Annales francaises d'anesthesie et de reanimation, 30(11), 848-850 (2011-10-08)
We are reporting the case of a bradykinin-mediated angioedema, secondary to the angiotensin converting enzyme inhibitors, which delayed treatment could have unfavorably influence the vital prognostic of the patient. Initially, the patient had an isolated edema of the superior lip.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service