Skip to Content
Merck
All Photos(3)

Key Documents

F7001

Sigma-Aldrich

5-Fluorotryptamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11FN2 · HCl
CAS Number:
Molecular Weight:
214.67
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

storage temp.

−20°C

Quality Level

SMILES string

Cl.NCCc1c[nH]c2ccc(F)cc12

InChI

1S/C10H11FN2.ClH/c11-8-1-2-10-9(5-8)7(3-4-12)6-13-10;/h1-2,5-6,13H,3-4,12H2;1H

InChI key

YZAWADGJXKCLTI-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thrombin: active-site topography.
L J Berliner et al.
Annals of the New York Academy of Sciences, 485, 80-95 (1986-01-01)
J Sastre Toraño et al.
Biomedical chromatography : BMC, 14(5), 306-310 (2000-08-29)
A validated new and precise reversed-phase high-performance liquid chromatographic method for the determination of melatonin in human plasma and cerebrospinal fluid, with 5-fluorotryptamine as internal standard, is described. Liquid-liquid extraction with dichloromethane was performed under alkaline conditions. After evaporation of
E A Tembe et al.
Parasitology, 106 ( Pt 1), 67-73 (1993-01-01)
5-HT is a candidate for the excitatory transmitter at the neuromuscular junction in trematodes including Fasciola hepatica. This study has determined the response of a muscle strip preparation from Fasciola hepatica to 5-HT and a range of agonists that distinguish
P A Mirau et al.
Biochemistry, 21(4), 615-620 (1982-02-16)
Fluorine-19 nuclear magnetic resonance (19F NMR), optical absorption, and fluorescence spectroscopy have been used to study the interaction of 5-fluorotryptamine (5FTA) with polynucleotides as a model for protein--nucleic acid interactions. In the presence of DNA, denatured DNA, poly(A), and poly(A).poly(U)
Kiowa S Bower et al.
European journal of pharmacology, 580(3), 291-297 (2007-12-18)
Antagonists, but not agonists, of the 5-HT3 receptor are useful therapeutic agents, and it is possible that partial agonists may also be potentially useful in the clinic. Here we show that 5-fluorotryptamine (5-FT) is a partial agonist at both 5-HT3A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service