Skip to Content
Merck
All Photos(3)

Key Documents

F6777

Sigma-Aldrich

Flecainide acetate salt

≥98% (TLC), powder, sodium channel blocker

Synonym(s):

N-(2-Piperidylmethyl)-2,5-bis-(2,2,2-trifluoroethoxy)benzamide acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20F6N2O3 · C2H4O2
CAS Number:
Molecular Weight:
474.39
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Flecainide acetate salt,

Quality Level

originator

Medicis

storage temp.

2-8°C

SMILES string

CC(O)=O.FC(F)(F)COc1ccc(OCC(F)(F)F)c(c1)C(=O)NCC2CCCCN2

InChI

1S/C17H20F6N2O3.C2H4O2/c18-16(19,20)9-27-12-4-5-14(28-10-17(21,22)23)13(7-12)15(26)25-8-11-3-1-2-6-24-11;1-2(3)4/h4-5,7,11,24H,1-3,6,8-10H2,(H,25,26);1H3,(H,3,4)

InChI key

RKXNZRPQSOPPRN-UHFFFAOYSA-N

Gene Information

human ... SCN5A(6331)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Class IC antiarrhythmic agent; sodium channel blocker

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Medicis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Somani
Clinical pharmacology and therapeutics, 27(4), 464-470 (1980-04-01)
A new class I antiarrhythmic, flecainide, was investigated in 10 patients to assess short-term efficacy and safety. All patients were hospitalized for 3 days; no antiarrhythmics were given on days 1 and 3. On day 2 flecainide 1 mg/kg was
Drug therapy. Flecainide.
D M Roden et al.
The New England journal of medicine, 315(1), 36-41 (1986-07-03)
Kosuke Doki et al.
Pharmacogenetics and genomics, 22(11), 777-783 (2012-09-04)
To investigate the association between age-related decline in flecainide clearance and CYP2D6 genotype, we conducted a population pharmacokinetic analysis of flecainide using routine therapeutic drug monitoring data. Population pharmacokinetic analysis was performed on retrospective data from 163 genotyped patients treated
P Maury et al.
Archives des maladies du coeur et des vaisseaux, 92(2), 273-277 (1999-03-17)
Poisoning with flecainide acetate is rare and associated with a high mortality. This usually occurs after massive ingestion but can also be observed during therapeutic overdose in patients with renal failure or with amiodarone therapy. The prognostic depends on the
J C Perry et al.
American heart journal, 124(6), 1614-1621 (1992-12-11)
A review of all published experience with flecainide in infants, children, and fetuses was performed to evaluate the appropriate place of the drug in pediatric practice and to determine dosing guidelines. A total of 704 case references was generated. Flecainide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service