Skip to Content
MilliporeSigma

Skip To

F4512

Anti-Human IgG (whole molecule)−FITC antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Rabbit Anti-Human IgG (whole molecule)−Fluorescein isothiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
$222.00
2 mL
Please contact Customer Service for Availability
$387.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF
Citations:
12

$222.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunofluorescence: 1:64-1:128 using Hep2 cells

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgG antibody plays a crucial role in humoral immune responses such as complement activation, phagocytosis, placental transport and cell surface-receptor binding. FITC (fluorescein isothiocyanate), a fluorochrome dye conjugation is useful in localization of antigen-antibody interaction of biological structures like cells or tissues. Anti-Human IgG (whole molecule) −FITC antibody diluted 1:100 in wash buffer is used in flow cytometry to analyze antibody affinity and virus binding. This antibody can also be used in indirect immunofluorescence antibody test. FITC labeled anti-human IgG antibody reacts specifically with human.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. Anti-Human IgG (whole molecule)−FITC antibody is specific for human IgG. Rabbit Anti-Human IgG is conjugated to crystalline fluorescein isothiocyanate (FITC) in an alkaline reaction and then further purified to remove free FITC.

Immunogen

IgG from pooled normal human serum

Application

Anti-Human IgG (whole molecule) −FITC antibody may be used as secondary antibody in indirect immunofluorescence and is also used in flow cytometry .
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
The antibody may be used for immunofluorescent staining of human peripheral blood lymphocytes at a working dilution of 1:64. A minimum working dilution of 1:64 was used on acetone fixed mouse liver cells and anti-nuclear antibody positive serum as the primary antibody. For flow cytometry to label NIH3T3 cells, the antibody was used at a working dilution of 1:100. The antibody was also used in ELISA using human sera.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F4637F5137F3512
conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

goat

technique(s)

immunofluorescence: 1:64-1:128 using Hep2 cells

technique(s)

ANA-indirect immunofluorescence: 1:128, direct immunofluorescence: 1:80

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100, indirect immunofluorescence: 1:100

technique(s)

immunofluorescence: 1:32-1:64 using Hep2 cells

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters