Key Documents
EP0030124928
Eppendorf® FAST PCR-Tube Strips
caps domed, pack of 120 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
cap (plug seal)
colorless cap
colorless tube
conical bottom
feature
PCR clean
graduations: no
neck thread Push fit
caps domed
packaging
pack of 120 ea
manufacturer/tradename
Eppendorf® 0030124928
technique(s)
PCR: suitable
capacity
0.1 mL
General description
  • Ideal for use with Fast cyclers such as the Mastercycler X50s and Fast enzymes.
  • 8 reaction tubes in strip-format − ideal for small sample volumes.
  • Easily adaptable for automation.
  • Sealable using domed strip-lids.
  • Certified free from human DNA, DNase, RNase and PCR inhibitors.
The Eppendorf Fast PCR Tube Strips made of Polyethylene transfer heat faster than the tubes made of traditional plastics. The Fast PCR Tube Strips are particularly suited to scientists using Fast PCR Reagent Kits, scientists trying to speed up protocols or cyclers with high heating and cooling rates - such as the Eppendorf Mastercycler X50 family.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG