Skip to Content
Merck
All Photos(1)

Key Documents

EMU016141

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Foxm1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCGTTAAGCAGGAACTGGAAGAGAAGGAGAATTGTCACCTGGAGCAGAATCGGGTTAAGGTTGAGGAGCCCTCAGGAGTGTCAACATCTTGGCAGGACTCTGTGTCTGAGAGGCCACCCTACTCTTATATGGCCATGATACAGTTTGCCATCAACAGCACTGAGAGAAAGCGCATGACCTTGAAGGACATCTACACTTGGATTGAGGACCACTTCCCTTACTTTAAGCACATTGCCAAGCCAGGCTGGAAGAACTCTATTCGTCACAACCTTTCTCTCCATGACATGTTTGTTCGAGAGACATCTGCCAATGGCAAGGTCTCCTTCTGGACCATTCACCCAAGTGCCAATCGCTACTTGACATTGGACCAAGTGTTTAAGCCACTGGAACCAGGGTCTCCACAATCGCCCGAGCACTTGGAATCACAGCA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxiao Li et al.
Journal of translational medicine, 11, 204-204 (2013-09-06)
Forkhead box transcription factor 1 (FOXM1) has been reported to overexpress and correlate with pathogenesis in a variety of human malignancies. However, little research has been done to investigate its clinical significance in gastric cancer. We examined the expression of
KanKan Yang et al.
Journal of experimental & clinical cancer research : CR, 34, 40-40 (2015-05-04)
The Forkhead box M1 (FOXM1) is an oncogenic transcription factor and plays a significant role in cell EMT, proliferation, metastasis in a multitude of human solid tumors including colorectal cancer (CRC). However, the underlying molecular mechanisms by which FoxM1 contributes
Satoru Inoguchi et al.
FEBS letters, 588(17), 3170-3179 (2014-07-08)
Here, we found that microRNA-24-1 (miR-24-1) is significantly reduced in bladder cancer (BC) tissues, suggesting that it functions as a tumour suppressor. Restoration of mature miR-24-1 inhibits cancer cell proliferation and induces apoptosis. Forkhead box protein M1 (FOXM1) is a
Jiujie Cui et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(10), 2595-2606 (2014-03-19)
The transcription factor Forkhead box protein M1 (FOXM1) plays critical roles in cancer development and progression. However, the regulatory role and underlying mechanisms of FOXM1 in cancer metabolism are unknown. In this study, we characterized the regulation of aerobic glycolysis
Weihua Jiang et al.
International journal of clinical and experimental pathology, 8(6), 6756-6763 (2015-08-12)
The oncogenic transcription factor forkhead box protein M1 (FOXM1) plays critical roles in gastric cancer (GC) development and progression. However, the underlying mechanisms has not fully demonstrated. Lactate dehydrogenase A (LDHA) is widely overexpressed in a series of cancers and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service