Skip to Content
MilliporeSigma

Skip To

D7802

Daidzein

≥98% (TLC), powder, carbonic anhydrase (CA) inhibitor

Synonym(s):

4′,7-Dihydroxy­iso­flavone, 7-Hydroxy-3-(4-hydroxy­phenyl)-4H-1-benzo­pyran-4-one, 7-Hydroxy-3-(4-hydroxy­phenyl)­chromone, Isoaurostatin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
$164.00
100 mg
Please contact Customer Service for Availability
$543.00

About This Item

Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
EC Number:
207-635-4
MDL number:
Beilstein/REAXYS Number:
231523
Assay:
≥98%
Form:
powder

$164.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Daidzein, ≥98%, synthetic

biological source

synthetic

Quality Segment

assay

≥98%

form

powder

mp

>290 °C

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=COc3cc(O)ccc3C2=O

InChI

1S/C15H10O4/c16-10-3-1-9(2-4-10)13-8-19-14-7-11(17)5-6-12(14)15(13)18/h1-8,16-17H

InChI key

ZQSIJRDFPHDXIC-UHFFFAOYSA-N

General description

Daidzein is an isoflavone present in soy. It regulates oxidative stress, angiogenesis, cell growth and apoptosis. Daidzein has antioxidant, antiestrogenic and antiatherogenic properties. It provides protection against osteoporosis.

Application

Daidzein has been used:
  • as a control to evaluate fertility in rats
  • to study its effects on ovarian cancer
  • to simulate in vitro antioxidant activity

Biochem/physiol Actions

Daidzein is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers.[1] Arrests cell cycle at G1 in Swiss 3T3 cells.[2]
Soy isoflavone daidzein protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

Packaging

Packaged under Argon.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
251600PHL891811162421
form

powder

form

solid

form

powder

form

-

assay

≥98%

assay

≥98% (HPLC)

assay

≥95.0% (HPLC)

assay

-

Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: 10 mg/mL

solubility

DMSO: 200 mg/mL

solubility

-

solubility

-

biological source

synthetic

biological source

-

biological source

-

biological source

-


Still not finding the right product?

Explore all of our products under Daidzein


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters